Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89102403-89102664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023020.1 XM_017023021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704433 c.709G>A p.V237M Chr.16: 89102646-89102646 on Build GRCh38
6592553 c.715C>A p.L239I Chr.16: 89102652-89102652 on Build GRCh38
6873111 c.703G>A p.D235N Chr.16: 89102640-89102640 on Build GRCh38
4063546 c.713T>C p.I238T Chr.16: 89102650-89102650 on Build GRCh38
6259713 c.721G>T p.V241L Chr.16: 89102658-89102658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377690227 Chr.16: 89102428-89102428 on Build GRCh38
rs759831732 Chr.16: 89102576-89102577 on Build GRCh38
rs150897718 Chr.16: 89102563-89102563 on Build GRCh38
rs576801165 Chr.16: 89102528-89102528 on Build GRCh38
rs763367217 Chr.16: 89102638-89102638 on Build GRCh38
rs775624630 Chr.16: 89102608-89102608 on Build GRCh38
rs371402253 Chr.16: 89102562-89102562 on Build GRCh38
rs113038318 Chr.16: 89102545-89102545 on Build GRCh38
rs1032476142 Chr.16: 89102623-89102623 on Build GRCh38
rs150374081 Chr.16: 89102609-89102609 on Build GRCh38
rs560458501 Chr.16: 89102585-89102585 on Build GRCh38
rs891108481 Chr.16: 89102601-89102601 on Build GRCh38
rs1025003738 Chr.16: 89102578-89102578 on Build GRCh38
rs1018421597 Chr.16: 89102481-89102481 on Build GRCh38
rs758951320 Chr.16: 89102580-89102580 on Build GRCh38
rs369865917 Chr.16: 89102595-89102595 on Build GRCh38
rs536459051 Chr.16: 89102477-89102477 on Build GRCh38
rs770578916 Chr.16: 89102597-89102597 on Build GRCh38
rs751901918 Chr.16: 89102564-89102565 on Build GRCh38
rs750797204 Chr.16: 89102630-89102630 on Build GRCh38
rs917560621 Chr.16: 89102538-89102538 on Build GRCh38
rs375555298 Chr.16: 89102590-89102590 on Build GRCh38
rs745722909 Chr.16: 89102604-89102604 on Build GRCh38
rs979238832 Chr.16: 89102502-89102502 on Build GRCh38
rs766852572 Chr.16: 89102642-89102642 on Build GRCh38
rs1018473735 Chr.16: 89102607-89102607 on Build GRCh38
rs778778276 Chr.16: 89102603-89102603 on Build GRCh38
rs904649168 Chr.16: 89102541-89102541 on Build GRCh38
rs777550478 Chr.16: 89102438-89102438 on Build GRCh38
rs987309154 Chr.16: 89102641-89102641 on Build GRCh38
rs765656614 Chr.16: 89102570-89102570 on Build GRCh38
rs762052567 Chr.16: 89102565-89102565 on Build GRCh38
rs956442814 Chr.16: 89102520-89102520 on Build GRCh38
rs373322510 Chr.16: 89102602-89102602 on Build GRCh38
rs763619768 Chr.16: 89102646-89102646 on Build GRCh38
rs1025611077 Chr.16: 89102431-89102431 on Build GRCh38
rs894651696 Chr.16: 89102564-89102564 on Build GRCh38
rs775714313 Chr.16: 89102556-89102556 on Build GRCh38
rs534255994 Chr.16: 89102514-89102514 on Build GRCh38
rs921778285 Chr.16: 89102497-89102497 on Build GRCh38
rs72817437 Chr.16: 89102592-89102592 on Build GRCh38
rs4782457 Chr.16: 89102573-89102573 on Build GRCh38
rs377215682 Chr.16: 89102533-89102533 on Build GRCh38
rs569063403 Chr.16: 89102439-89102439 on Build GRCh38
rs776677384 Chr.16: 89102622-89102622 on Build GRCh38
rs368638619 Chr.16: 89102555-89102555 on Build GRCh38
rs4782458 Chr.16: 89102527-89102527 on Build GRCh38
rs550710102 Chr.16: 89102437-89102437 on Build GRCh38
rs145583876 Chr.16: 89102626-89102626 on Build GRCh38
rs752104014 Chr.16: 89102643-89102643 on Build GRCh38
rs764828984 Chr.16: 89102560-89102561 on Build GRCh38
rs7200676 Chr.16: 89102503-89102503 on Build GRCh38
rs113197046 Chr.16: 89102540-89102540 on Build GRCh38
rs113568544 Chr.16: 89102493-89102493 on Build GRCh38
rs963094556 Chr.16: 89102456-89102456 on Build GRCh38
rs898538145 Chr.16: 89102424-89102424 on Build GRCh38
rs567213963 Chr.16: 89102551-89102551 on Build GRCh38
rs765454020 Chr.16: 89102628-89102628 on Build GRCh38
rs775694971 Chr.16: 89102610-89102610 on Build GRCh38
rs907664130 Chr.16: 89102451-89102451 on Build GRCh38
rs891392859 Chr.16: 89102447-89102447 on Build GRCh38
rs772127402 Chr.16: 89102606-89102606 on Build GRCh38
rs139367843 Chr.16: 89102591-89102591 on Build GRCh38
rs544290058 Chr.16: 89102534-89102534 on Build GRCh38
rs189491917 Chr.16: 89102579-89102579 on Build GRCh38
rs550622194 Chr.16: 89102645-89102645 on Build GRCh38

View Full Product Details