Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49067294-49067561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC120
Entrez Gene ID
90060
Gene Name
coiled-coil domain containing 120
Gene Aliases
JM11
UniGene
Hs.522643
Gene Chromosome Location
Chr.X: 49053432-49069858 on Build GRCh38
Transcripts
NM_001163321.2 NM_001163322.2 NM_001163323.2 NM_001271835.1 NM_001271836.1 NM_033626.3

SNP Detail

SNP ID Chromosome Location
rs782271112 Chr.X: 49067332-49067332 on Build GRCh38
rs781787357 Chr.X: 49067522-49067522 on Build GRCh38
rs782534366 Chr.X: 49067430-49067430 on Build GRCh38
rs370154494 Chr.X: 49067350-49067350 on Build GRCh38
rs782780443 Chr.X: 49067326-49067326 on Build GRCh38
rs782116740 Chr.X: 49067373-49067373 on Build GRCh38
rs782098187 Chr.X: 49067483-49067483 on Build GRCh38
rs55654138 Chr.X: 49067360-49067360 on Build GRCh38
rs781829861 Chr.X: 49067516-49067517 on Build GRCh38
rs782779794 Chr.X: 49067321-49067321 on Build GRCh38
rs782619912 Chr.X: 49067520-49067520 on Build GRCh38
rs782058115 Chr.X: 49067511-49067511 on Build GRCh38
rs781826924 Chr.X: 49067517-49067517 on Build GRCh38
rs781798809 Chr.X: 49067327-49067327 on Build GRCh38
rs782180094 Chr.X: 49067529-49067529 on Build GRCh38
rs782001015 Chr.X: 49067371-49067371 on Build GRCh38
rs782281432 Chr.X: 49067339-49067339 on Build GRCh38
rs782760184 Chr.X: 49067432-49067432 on Build GRCh38
rs782584415 Chr.X: 49067525-49067525 on Build GRCh38
rs781922483 Chr.X: 49067351-49067351 on Build GRCh38
rs782512531 Chr.X: 49067333-49067333 on Build GRCh38
rs782186978 Chr.X: 49067458-49067458 on Build GRCh38
rs782700988 Chr.X: 49067418-49067418 on Build GRCh38
rs781816554 Chr.X: 49067510-49067510 on Build GRCh38
rs898949465 Chr.X: 49067361-49067361 on Build GRCh38
rs201466297 Chr.X: 49067501-49067501 on Build GRCh38
rs782425279 Chr.X: 49067425-49067425 on Build GRCh38
rs782160876 Chr.X: 49067491-49067491 on Build GRCh38
rs781919882 Chr.X: 49067481-49067481 on Build GRCh38
rs374295188 Chr.X: 49067479-49067479 on Build GRCh38
rs782804086 Chr.X: 49067380-49067380 on Build GRCh38
rs200874918 Chr.X: 49067443-49067443 on Build GRCh38
rs782537429 Chr.X: 49067447-49067447 on Build GRCh38
rs782482732 Chr.X: 49067461-49067461 on Build GRCh38
rs782469793 Chr.X: 49067417-49067417 on Build GRCh38
rs911802103 Chr.X: 49067349-49067349 on Build GRCh38
rs781988396 Chr.X: 49067490-49067490 on Build GRCh38
rs782334530 Chr.X: 49067363-49067363 on Build GRCh38
rs782605460 Chr.X: 49067463-49067463 on Build GRCh38
rs782428766 Chr.X: 49067328-49067328 on Build GRCh38
rs782255200 Chr.X: 49067474-49067474 on Build GRCh38
rs782011731 Chr.X: 49067385-49067385 on Build GRCh38
rs371323348 Chr.X: 49067477-49067477 on Build GRCh38
rs782528230 Chr.X: 49067523-49067523 on Build GRCh38
rs959629328 Chr.X: 49067454-49067454 on Build GRCh38
rs782774222 Chr.X: 49067487-49067487 on Build GRCh38
rs782586777 Chr.X: 49067448-49067448 on Build GRCh38

View Full Product Details