Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43362029-43362262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD177
Entrez Gene ID
57126
Gene Name
CD177 molecule
Gene Aliases
HNA-2a, HNA2A, NB1, NB1 GP, PRV-1, PRV1
UniGene
Hs.232165
Gene Chromosome Location
Chr.19: 43353659-43366075 on Build GRCh38
Transcripts
NM_020406.3 XM_017027021.1 XM_017027022.1

SNP Detail

SNP ID Chromosome Location
rs760918291 Chr.19: 43362175-43362175 on Build GRCh38
rs760013413 Chr.19: 43362167-43362167 on Build GRCh38
rs767588126 Chr.19: 43362106-43362106 on Build GRCh38
rs754479368 Chr.19: 43362174-43362174 on Build GRCh38
rs773632781 Chr.19: 43362087-43362087 on Build GRCh38
rs185102629 Chr.19: 43362103-43362103 on Build GRCh38
rs199770370 Chr.19: 43362182-43362182 on Build GRCh38
rs766186727 Chr.19: 43362079-43362080 on Build GRCh38
rs747793913 Chr.19: 43362160-43362160 on Build GRCh38
rs180771874 Chr.19: 43362096-43362096 on Build GRCh38
rs761501813 Chr.19: 43362070-43362070 on Build GRCh38
rs1001220105 Chr.19: 43362242-43362242 on Build GRCh38
rs776964743 Chr.19: 43362221-43362221 on Build GRCh38
rs550497658 Chr.19: 43362195-43362195 on Build GRCh38
rs558473917 Chr.19: 43362107-43362107 on Build GRCh38
rs751529556 Chr.19: 43362099-43362099 on Build GRCh38
rs753512453 Chr.19: 43362172-43362174 on Build GRCh38
rs201285428 Chr.19: 43362122-43362122 on Build GRCh38
rs758340894 Chr.19: 43362123-43362123 on Build GRCh38
rs780740088 Chr.19: 43362184-43362184 on Build GRCh38
rs771307102 Chr.19: 43362193-43362193 on Build GRCh38
rs752153110 Chr.19: 43362238-43362238 on Build GRCh38
rs772517528 Chr.19: 43362196-43362196 on Build GRCh38
rs752895773 Chr.19: 43362170-43362170 on Build GRCh38
rs780068747 Chr.19: 43362129-43362129 on Build GRCh38
rs369897962 Chr.19: 43362192-43362192 on Build GRCh38
rs772190359 Chr.19: 43362049-43362049 on Build GRCh38
rs559438445 Chr.19: 43362162-43362162 on Build GRCh38
rs369478901 Chr.19: 43362168-43362168 on Build GRCh38
rs541810499 Chr.19: 43362181-43362181 on Build GRCh38
rs767369105 Chr.19: 43362235-43362235 on Build GRCh38
rs768188920 Chr.19: 43362148-43362148 on Build GRCh38
rs375865849 Chr.19: 43362146-43362146 on Build GRCh38
rs778439043 Chr.19: 43362204-43362205 on Build GRCh38
rs201215682 Chr.19: 43362183-43362183 on Build GRCh38
rs763438044 Chr.19: 43362089-43362089 on Build GRCh38
rs750200960 Chr.19: 43362180-43362180 on Build GRCh38
rs770112355 Chr.19: 43362071-43362071 on Build GRCh38
rs764374704 Chr.19: 43362220-43362220 on Build GRCh38
rs761991629 Chr.19: 43362222-43362222 on Build GRCh38
rs780606678 Chr.19: 43362157-43362157 on Build GRCh38
rs754878660 Chr.19: 43362232-43362232 on Build GRCh38
rs746530573 Chr.19: 43362056-43362056 on Build GRCh38
rs376877003 Chr.19: 43362165-43362165 on Build GRCh38
rs760863240 Chr.19: 43362205-43362205 on Build GRCh38
rs775657244 Chr.19: 43362199-43362199 on Build GRCh38
rs763313723 Chr.19: 43362164-43362164 on Build GRCh38
rs540826369 Chr.19: 43362154-43362154 on Build GRCh38
rs779369220 Chr.19: 43362055-43362055 on Build GRCh38
rs755697585 Chr.19: 43362189-43362189 on Build GRCh38
rs776366973 Chr.19: 43362069-43362069 on Build GRCh38
rs748050266 Chr.19: 43362231-43362231 on Build GRCh38
rs1039073957 Chr.19: 43362133-43362133 on Build GRCh38
rs766185106 Chr.19: 43362224-43362224 on Build GRCh38
rs768150769 Chr.19: 43362058-43362058 on Build GRCh38
rs774946620 Chr.19: 43362166-43362166 on Build GRCh38
rs746453817 Chr.19: 43362144-43362144 on Build GRCh38
rs757686752 Chr.19: 43362055-43362070 on Build GRCh38
rs528133050 Chr.19: 43362177-43362177 on Build GRCh38
rs751378408 Chr.19: 43362228-43362228 on Build GRCh38
rs756448661 Chr.19: 43362117-43362117 on Build GRCh38

View Full Product Details