Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129250577-129250840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634068 c.79G>A p.V27I Chr.3: 129250723-129250723 on Build GRCh38
1202069 c.90+2C>A p.? Chr.3: 129250736-129250736 on Build GRCh38
338318 c.77C>T p.A26V Chr.3: 129250721-129250721 on Build GRCh38
1166880 c.50A>G p.N17S Chr.3: 129250694-129250694 on Build GRCh38
1632797 c.66T>C p.L22L Chr.3: 129250710-129250710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776645124 Chr.3: 129250806-129250806 on Build GRCh38
rs527946344 Chr.3: 129250788-129250788 on Build GRCh38
rs922144335 Chr.3: 129250790-129250790 on Build GRCh38
rs772597812 Chr.3: 129250782-129250782 on Build GRCh38
rs779406263 Chr.3: 129250769-129250769 on Build GRCh38
rs753752221 Chr.3: 129250643-129250643 on Build GRCh38
rs745671291 Chr.3: 129250616-129250616 on Build GRCh38
rs766381465 Chr.3: 129250639-129250639 on Build GRCh38
rs748374022 Chr.3: 129250687-129250687 on Build GRCh38
rs760769785 Chr.3: 129250695-129250695 on Build GRCh38
rs984842211 Chr.3: 129250815-129250815 on Build GRCh38
rs756545541 Chr.3: 129250671-129250671 on Build GRCh38
rs748570330 Chr.3: 129250777-129250777 on Build GRCh38
rs768366396 Chr.3: 129250805-129250805 on Build GRCh38
rs765031291 Chr.3: 129250659-129250659 on Build GRCh38
rs567842445 Chr.3: 129250810-129250810 on Build GRCh38
rs996353483 Chr.3: 129250628-129250628 on Build GRCh38
rs370174152 Chr.3: 129250756-129250756 on Build GRCh38
rs968239276 Chr.3: 129250654-129250654 on Build GRCh38
rs199598040 Chr.3: 129250736-129250736 on Build GRCh38
rs144886799 Chr.3: 129250802-129250802 on Build GRCh38
rs774859705 Chr.3: 129250682-129250682 on Build GRCh38
rs1022432771 Chr.3: 129250646-129250646 on Build GRCh38
rs973882890 Chr.3: 129250780-129250780 on Build GRCh38
rs372871313 Chr.3: 129250708-129250708 on Build GRCh38
rs749778479 Chr.3: 129250678-129250678 on Build GRCh38
rs752549568 Chr.3: 129250751-129250751 on Build GRCh38
rs754754111 Chr.3: 129250652-129250652 on Build GRCh38
rs79824567 Chr.3: 129250668-129250668 on Build GRCh38
rs773136279 Chr.3: 129250694-129250694 on Build GRCh38
rs538081107 Chr.3: 129250813-129250813 on Build GRCh38
rs376876037 Chr.3: 129250753-129250753 on Build GRCh38
rs373204048 Chr.3: 129250773-129250773 on Build GRCh38
rs765016885 Chr.3: 129250721-129250721 on Build GRCh38
rs752283936 Chr.3: 129250662-129250662 on Build GRCh38
rs759400182 Chr.3: 129250712-129250712 on Build GRCh38
rs369091902 Chr.3: 129250690-129250690 on Build GRCh38
rs550232110 Chr.3: 129250814-129250814 on Build GRCh38
rs780566347 Chr.3: 129250677-129250677 on Build GRCh38
rs760515992 Chr.3: 129250635-129250635 on Build GRCh38
rs900725386 Chr.3: 129250627-129250627 on Build GRCh38
rs754399088 Chr.3: 129250768-129250769 on Build GRCh38
rs766392519 Chr.3: 129250696-129250696 on Build GRCh38
rs368427024 Chr.3: 129250754-129250754 on Build GRCh38
rs755608748 Chr.3: 129250765-129250765 on Build GRCh38
rs758115690 Chr.3: 129250665-129250665 on Build GRCh38

View Full Product Details