Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136823388-136823661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL6
Entrez Gene ID
55684
Gene Name
RAB, member RAS oncogene family-like 6
Gene Aliases
C9orf86, PARF, RBEL1, pp8875
UniGene
Hs.370555
Gene Chromosome Location
Chr.9: 136807922-136841187 on Build GRCh38
Transcripts
NM_001173988.1 NM_001173989.2 NM_024718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5835678 c.235_236insG p.Q79fs*14 Chr.9: 136823629-136823630 on Build GRCh38
4478212 c.455C>T p.P152L Chr.9: 136823615-136823615 on Build GRCh38
5835672 c.234_235insA p.Q79fs*14 Chr.9: 136823628-136823629 on Build GRCh38
4534787 c.445G>A p.E149K Chr.9: 136823605-136823605 on Build GRCh38
3655841 c.453C>T p.I151I Chr.9: 136823613-136823613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375365327 Chr.9: 136823487-136823487 on Build GRCh38
rs962827295 Chr.9: 136823579-136823579 on Build GRCh38
rs532298031 Chr.9: 136823432-136823432 on Build GRCh38
rs115051707 Chr.9: 136823488-136823488 on Build GRCh38
rs938996711 Chr.9: 136823458-136823458 on Build GRCh38
rs766655068 Chr.9: 136823636-136823636 on Build GRCh38
rs774661349 Chr.9: 136823538-136823538 on Build GRCh38
rs756553824 Chr.9: 136823568-136823568 on Build GRCh38
rs762061814 Chr.9: 136823481-136823481 on Build GRCh38
rs548702471 Chr.9: 136823556-136823556 on Build GRCh38
rs74416852 Chr.9: 136823529-136823529 on Build GRCh38
rs149867930 Chr.9: 136823598-136823598 on Build GRCh38
rs773325201 Chr.9: 136823628-136823628 on Build GRCh38
rs988309923 Chr.9: 136823456-136823456 on Build GRCh38
rs770953163 Chr.9: 136823517-136823517 on Build GRCh38
rs769861226 Chr.9: 136823482-136823482 on Build GRCh38
rs774636565 Chr.9: 136823601-136823601 on Build GRCh38
rs1046372407 Chr.9: 136823441-136823441 on Build GRCh38
rs763090414 Chr.9: 136823525-136823525 on Build GRCh38
rs758711900 Chr.9: 136823502-136823503 on Build GRCh38
rs768779181 Chr.9: 136823417-136823417 on Build GRCh38
rs761059018 Chr.9: 136823632-136823632 on Build GRCh38
rs778407287 Chr.9: 136823586-136823586 on Build GRCh38
rs767941580 Chr.9: 136823476-136823476 on Build GRCh38
rs371600377 Chr.9: 136823521-136823521 on Build GRCh38
rs767763981 Chr.9: 136823562-136823562 on Build GRCh38
rs747803921 Chr.9: 136823498-136823498 on Build GRCh38
rs745817863 Chr.9: 136823599-136823599 on Build GRCh38
rs747795737 Chr.9: 136823590-136823590 on Build GRCh38
rs368455449 Chr.9: 136823578-136823578 on Build GRCh38
rs746771659 Chr.9: 136823594-136823594 on Build GRCh38
rs748842936 Chr.9: 136823486-136823486 on Build GRCh38
rs368002409 Chr.9: 136823595-136823595 on Build GRCh38
rs766859620 Chr.9: 136823571-136823571 on Build GRCh38
rs114441485 Chr.9: 136823483-136823483 on Build GRCh38
rs762092077 Chr.9: 136823602-136823602 on Build GRCh38
rs981762428 Chr.9: 136823472-136823472 on Build GRCh38
rs771796482 Chr.9: 136823508-136823508 on Build GRCh38
rs758110733 Chr.9: 136823591-136823591 on Build GRCh38
rs34054033 Chr.9: 136823446-136823447 on Build GRCh38
rs371702474 Chr.9: 136823580-136823580 on Build GRCh38
rs750837741 Chr.9: 136823567-136823567 on Build GRCh38
rs913629344 Chr.9: 136823460-136823460 on Build GRCh38
rs746889543 Chr.9: 136823516-136823516 on Build GRCh38
rs577864527 Chr.9: 136823581-136823581 on Build GRCh38
rs376408551 Chr.9: 136823592-136823592 on Build GRCh38
rs957862554 Chr.9: 136823433-136823433 on Build GRCh38
rs773126745 Chr.9: 136823511-136823511 on Build GRCh38
rs767826857 Chr.9: 136823606-136823606 on Build GRCh38
rs774388602 Chr.9: 136823467-136823467 on Build GRCh38

View Full Product Details