Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15436333-15436606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF1
Entrez Gene ID
85403
Gene Name
ELL associated factor 1
Gene Aliases
-
UniGene
Hs.716733
Gene Chromosome Location
Chr.3: 15427557-15442613 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1 XM_011534165.1 XM_011534166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580933 c.584C>T p.S195F Chr.3: 15436399-15436399 on Build GRCh38
5620732 c.757C>T p.L253F Chr.3: 15436572-15436572 on Build GRCh38
5660594 c.724C>T p.R242W Chr.3: 15436539-15436539 on Build GRCh38
1327791 c.548T>G p.I183S Chr.3: 15436363-15436363 on Build GRCh38
5751065 c.665C>T p.P222L Chr.3: 15436480-15436480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745886469 Chr.3: 15436454-15436454 on Build GRCh38
rs34668493 Chr.3: 15436433-15436433 on Build GRCh38
rs747229897 Chr.3: 15436580-15436580 on Build GRCh38
rs1039722210 Chr.3: 15436563-15436563 on Build GRCh38
rs772292952 Chr.3: 15436458-15436458 on Build GRCh38
rs145331815 Chr.3: 15436480-15436480 on Build GRCh38
rs367835501 Chr.3: 15436520-15436520 on Build GRCh38
rs542702975 Chr.3: 15436553-15436553 on Build GRCh38
rs964403754 Chr.3: 15436575-15436575 on Build GRCh38
rs752347537 Chr.3: 15436421-15436421 on Build GRCh38
rs549005284 Chr.3: 15436406-15436406 on Build GRCh38
rs375782986 Chr.3: 15436395-15436395 on Build GRCh38
rs747233190 Chr.3: 15436465-15436465 on Build GRCh38
rs149438944 Chr.3: 15436513-15436513 on Build GRCh38
rs758977481 Chr.3: 15436485-15436485 on Build GRCh38
rs777984333 Chr.3: 15436430-15436430 on Build GRCh38
rs759044498 Chr.3: 15436528-15436528 on Build GRCh38
rs772386193 Chr.3: 15436367-15436367 on Build GRCh38
rs146901213 Chr.3: 15436521-15436521 on Build GRCh38
rs895172697 Chr.3: 15436486-15436486 on Build GRCh38
rs199650019 Chr.3: 15436434-15436434 on Build GRCh38
rs757570655 Chr.3: 15436374-15436374 on Build GRCh38
rs772864991 Chr.3: 15436476-15436476 on Build GRCh38
rs200760056 Chr.3: 15436539-15436539 on Build GRCh38
rs144767041 Chr.3: 15436540-15436540 on Build GRCh38
rs535493318 Chr.3: 15436366-15436366 on Build GRCh38
rs766391671 Chr.3: 15436399-15436399 on Build GRCh38
rs775789874 Chr.3: 15436462-15436462 on Build GRCh38
rs754113318 Chr.3: 15436514-15436514 on Build GRCh38
rs1015920075 Chr.3: 15436549-15436549 on Build GRCh38
rs578172246 Chr.3: 15436499-15436499 on Build GRCh38
rs761644298 Chr.3: 15436386-15436386 on Build GRCh38
rs145737507 Chr.3: 15436477-15436477 on Build GRCh38
rs774077299 Chr.3: 15436572-15436572 on Build GRCh38
rs780270918 Chr.3: 15436529-15436529 on Build GRCh38
rs781495879 Chr.3: 15436548-15436548 on Build GRCh38
rs1012770914 Chr.3: 15436512-15436512 on Build GRCh38
rs755830257 Chr.3: 15436426-15436426 on Build GRCh38
rs114187957 Chr.3: 15436442-15436442 on Build GRCh38
rs767094068 Chr.3: 15436420-15436420 on Build GRCh38
rs770761478 Chr.3: 15436570-15436570 on Build GRCh38
rs982140519 Chr.3: 15436404-15436404 on Build GRCh38
rs762788329 Chr.3: 15436390-15436390 on Build GRCh38
rs769060506 Chr.3: 15436474-15436474 on Build GRCh38
rs754503030 Chr.3: 15436408-15436408 on Build GRCh38
rs765132915 Chr.3: 15436387-15436387 on Build GRCh38
rs756050008 Chr.3: 15436384-15436384 on Build GRCh38
rs752261344 Chr.3: 15436505-15436505 on Build GRCh38
rs201575159 Chr.3: 15436481-15436481 on Build GRCh38
rs775877887 Chr.3: 15436370-15436370 on Build GRCh38
rs757663336 Chr.3: 15436516-15436516 on Build GRCh38
rs950692079 Chr.3: 15436355-15436355 on Build GRCh38
rs17854099 Chr.3: 15436569-15436569 on Build GRCh38
rs908913215 Chr.3: 15436388-15436388 on Build GRCh38

View Full Product Details