Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182719864-182720129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
UniGene
Hs.516632
Gene Chromosome Location
Chr.2: 182716041-182780025 on Build GRCh38
Transcripts
NM_001271581.1 NM_018981.2 NR_073365.1 NR_073366.1 NR_073367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832124 c.316G>A p.D106N Chr.2: 182720118-182720118 on Build GRCh38
4410795 c.247G>A p.A83T Chr.2: 182720049-182720049 on Build GRCh38
6156398 c.277C>T p.R93W Chr.2: 182720079-182720079 on Build GRCh38
1494696 c.233T>A p.L78* Chr.2: 182720035-182720035 on Build GRCh38
3391333 c.210C>A p.N70K Chr.2: 182720012-182720012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6729801 Chr.2: 182720028-182720028 on Build GRCh38
rs904242919 Chr.2: 182719963-182719963 on Build GRCh38
rs575073487 Chr.2: 182719969-182719969 on Build GRCh38
rs756375421 Chr.2: 182720091-182720091 on Build GRCh38
rs748413503 Chr.2: 182719985-182719985 on Build GRCh38
rs1042137212 Chr.2: 182719934-182719934 on Build GRCh38
rs770906503 Chr.2: 182720015-182720015 on Build GRCh38
rs769071349 Chr.2: 182719966-182719966 on Build GRCh38
rs1002315771 Chr.2: 182720087-182720087 on Build GRCh38
rs969357396 Chr.2: 182720080-182720080 on Build GRCh38
rs983437541 Chr.2: 182720047-182720047 on Build GRCh38
rs1031549494 Chr.2: 182719998-182719998 on Build GRCh38
rs1003731559 Chr.2: 182719961-182719961 on Build GRCh38
rs781569732 Chr.2: 182720082-182720082 on Build GRCh38
rs769964845 Chr.2: 182719990-182719990 on Build GRCh38
rs772844876 Chr.2: 182719949-182719949 on Build GRCh38
rs895012054 Chr.2: 182720008-182720008 on Build GRCh38
rs150009047 Chr.2: 182720027-182720027 on Build GRCh38
rs774221574 Chr.2: 182720020-182720020 on Build GRCh38
rs200695145 Chr.2: 182720013-182720013 on Build GRCh38
rs767468170 Chr.2: 182720023-182720023 on Build GRCh38
rs376345220 Chr.2: 182719996-182719996 on Build GRCh38
rs932441972 Chr.2: 182719893-182719893 on Build GRCh38
rs749520487 Chr.2: 182720094-182720094 on Build GRCh38
rs748462996 Chr.2: 182720089-182720089 on Build GRCh38
rs906263835 Chr.2: 182719944-182719944 on Build GRCh38
rs764792929 Chr.2: 182720073-182720073 on Build GRCh38
rs374378803 Chr.2: 182719919-182719919 on Build GRCh38
rs1042775372 Chr.2: 182719940-182719940 on Build GRCh38
rs753399511 Chr.2: 182720050-182720050 on Build GRCh38
rs755450702 Chr.2: 182720079-182720079 on Build GRCh38
rs763602266 Chr.2: 182720044-182720044 on Build GRCh38
rs770971543 Chr.2: 182720099-182720099 on Build GRCh38
rs762853337 Chr.2: 182720024-182720024 on Build GRCh38
rs759532288 Chr.2: 182720021-182720021 on Build GRCh38
rs554825684 Chr.2: 182719937-182719937 on Build GRCh38
rs756788319 Chr.2: 182720051-182720051 on Build GRCh38
rs879822227 Chr.2: 182720062-182720062 on Build GRCh38

View Full Product Details