Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31464038-31464309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG3
Entrez Gene ID
1830
Gene Name
desmoglein 3
Gene Aliases
CDHF6, PVA
UniGene
Hs.1925
Gene Chromosome Location
Chr.18: 31447769-31478702 on Build GRCh38
Transcripts
NM_001944.2 XM_011525850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274886 c.1085G>A p.R362Q Chr.18: 31464196-31464196 on Build GRCh38
4071789 c.1011T>C p.Y337Y Chr.18: 31464122-31464122 on Build GRCh38
3717881 c.1178A>C p.Q393P Chr.18: 31464289-31464289 on Build GRCh38
5854072 c.1135G>A p.E379K Chr.18: 31464246-31464246 on Build GRCh38
1711165 c.1091G>A p.R364Q Chr.18: 31464202-31464202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771155268 Chr.18: 31464255-31464255 on Build GRCh38
rs113083364 Chr.18: 31464122-31464122 on Build GRCh38
rs778892043 Chr.18: 31464072-31464072 on Build GRCh38
rs893360840 Chr.18: 31464214-31464214 on Build GRCh38
rs943773773 Chr.18: 31464088-31464088 on Build GRCh38
rs779523046 Chr.18: 31464086-31464086 on Build GRCh38
rs144245283 Chr.18: 31464063-31464063 on Build GRCh38
rs776681686 Chr.18: 31464158-31464158 on Build GRCh38
rs781106006 Chr.18: 31464066-31464066 on Build GRCh38
rs537142426 Chr.18: 31464128-31464128 on Build GRCh38
rs767495340 Chr.18: 31464273-31464273 on Build GRCh38
rs377115190 Chr.18: 31464262-31464262 on Build GRCh38
rs750246751 Chr.18: 31464202-31464202 on Build GRCh38
rs778432093 Chr.18: 31464228-31464228 on Build GRCh38
rs142415015 Chr.18: 31464261-31464261 on Build GRCh38
rs146025709 Chr.18: 31464250-31464250 on Build GRCh38
rs369572434 Chr.18: 31464231-31464231 on Build GRCh38
rs141272063 Chr.18: 31464138-31464138 on Build GRCh38
rs555118940 Chr.18: 31464137-31464137 on Build GRCh38
rs755798646 Chr.18: 31464208-31464208 on Build GRCh38
rs754601362 Chr.18: 31464227-31464227 on Build GRCh38
rs763070535 Chr.18: 31464267-31464267 on Build GRCh38
rs750153030 Chr.18: 31464068-31464068 on Build GRCh38
rs779417216 Chr.18: 31464117-31464117 on Build GRCh38
rs372851025 Chr.18: 31464237-31464237 on Build GRCh38
rs779786641 Chr.18: 31464216-31464216 on Build GRCh38
rs376501967 Chr.18: 31464126-31464126 on Build GRCh38
rs73414212 Chr.18: 31464196-31464196 on Build GRCh38
rs369679425 Chr.18: 31464062-31464062 on Build GRCh38
rs762944000 Chr.18: 31464222-31464222 on Build GRCh38
rs376395413 Chr.18: 31464065-31464065 on Build GRCh38
rs576091748 Chr.18: 31464096-31464096 on Build GRCh38
rs772983108 Chr.18: 31464277-31464277 on Build GRCh38
rs753439250 Chr.18: 31464225-31464225 on Build GRCh38
rs747436564 Chr.18: 31464242-31464242 on Build GRCh38
rs768114247 Chr.18: 31464102-31464102 on Build GRCh38
rs755838960 Chr.18: 31464085-31464085 on Build GRCh38
rs767344775 Chr.18: 31464201-31464201 on Build GRCh38
rs540587123 Chr.18: 31464195-31464195 on Build GRCh38
rs747649462 Chr.18: 31464230-31464230 on Build GRCh38
rs772095495 Chr.18: 31464098-31464099 on Build GRCh38
rs748215668 Chr.18: 31464092-31464092 on Build GRCh38
rs775771731 Chr.18: 31464266-31464266 on Build GRCh38
rs761910376 Chr.18: 31464200-31464200 on Build GRCh38
rs1023071172 Chr.18: 31464236-31464236 on Build GRCh38
rs773689509 Chr.18: 31464131-31464131 on Build GRCh38
rs759714163 Chr.18: 31464177-31464177 on Build GRCh38
rs980425164 Chr.18: 31464145-31464145 on Build GRCh38
rs773454351 Chr.18: 31464131-31464132 on Build GRCh38
rs773725958 Chr.18: 31464148-31464148 on Build GRCh38
rs577490317 Chr.18: 31464256-31464256 on Build GRCh38

View Full Product Details