Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67306639-67306910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521914.2 XM_011521915.2 XM_011521916.2 XM_011521917.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

SNP Detail

SNP ID Chromosome Location
rs918697327 Chr.15: 67306668-67306668 on Build GRCh38
rs113683808 Chr.15: 67306807-67306807 on Build GRCh38
rs751304428 Chr.15: 67306671-67306672 on Build GRCh38
rs559474498 Chr.15: 67306814-67306814 on Build GRCh38
rs1037102788 Chr.15: 67306662-67306662 on Build GRCh38
rs759759022 Chr.15: 67306731-67306731 on Build GRCh38
rs762986680 Chr.15: 67306863-67306863 on Build GRCh38
rs988128581 Chr.15: 67306857-67306857 on Build GRCh38
rs933439019 Chr.15: 67306853-67306853 on Build GRCh38
rs770506141 Chr.15: 67306870-67306870 on Build GRCh38
rs1012131562 Chr.15: 67306774-67306774 on Build GRCh38
rs761483890 Chr.15: 67306779-67306779 on Build GRCh38
rs192916978 Chr.15: 67306667-67306667 on Build GRCh38
rs114714402 Chr.15: 67306837-67306837 on Build GRCh38
rs749909817 Chr.15: 67306815-67306815 on Build GRCh38
rs919458888 Chr.15: 67306844-67306844 on Build GRCh38
rs768005834 Chr.15: 67306869-67306869 on Build GRCh38
rs939192113 Chr.15: 67306671-67306671 on Build GRCh38
rs1056739995 Chr.15: 67306679-67306679 on Build GRCh38
rs557197604 Chr.15: 67306705-67306705 on Build GRCh38
rs895024107 Chr.15: 67306738-67306738 on Build GRCh38
rs573720661 Chr.15: 67306757-67306757 on Build GRCh38

View Full Product Details