Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1979192-1979417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
UniGene
Hs.651905
Gene Chromosome Location
Chr.19: 1941149-1981338 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259499.3 XM_005259500.1 XM_005259501.2 XM_017026297.1 XM_017026298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273677 c.776C>T p.T259M Chr.19: 1979326-1979326 on Build GRCh38
1202503 c.787C>T p.R263W Chr.19: 1979337-1979337 on Build GRCh38
6636800 c.779T>G p.L260R Chr.19: 1979329-1979329 on Build GRCh38
6212967 c.791A>G p.Y264C Chr.19: 1979341-1979341 on Build GRCh38
5513424 c.777G>A p.T259T Chr.19: 1979327-1979327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776152304 Chr.19: 1979252-1979252 on Build GRCh38
rs767521303 Chr.19: 1979284-1979284 on Build GRCh38
rs371407966 Chr.19: 1979314-1979314 on Build GRCh38
rs955699683 Chr.19: 1979304-1979304 on Build GRCh38
rs746355518 Chr.19: 1979233-1979233 on Build GRCh38
rs373599985 Chr.19: 1979221-1979221 on Build GRCh38
rs770798173 Chr.19: 1979354-1979354 on Build GRCh38
rs762868052 Chr.19: 1979264-1979264 on Build GRCh38
rs754133362 Chr.19: 1979372-1979372 on Build GRCh38
rs774317639 Chr.19: 1979355-1979355 on Build GRCh38
rs760867567 Chr.19: 1979367-1979367 on Build GRCh38
rs772975375 Chr.19: 1979336-1979336 on Build GRCh38
rs754934394 Chr.19: 1979283-1979283 on Build GRCh38
rs137921162 Chr.19: 1979396-1979396 on Build GRCh38
rs752677240 Chr.19: 1979366-1979366 on Build GRCh38
rs142134343 Chr.19: 1979327-1979327 on Build GRCh38
rs749457852 Chr.19: 1979296-1979296 on Build GRCh38
rs750798326 Chr.19: 1979391-1979391 on Build GRCh38
rs751596129 Chr.19: 1979282-1979282 on Build GRCh38
rs762490233 Chr.19: 1979351-1979351 on Build GRCh38
rs368618107 Chr.19: 1979364-1979364 on Build GRCh38
rs934172875 Chr.19: 1979254-1979254 on Build GRCh38
rs764952388 Chr.19: 1979262-1979262 on Build GRCh38
rs747657577 Chr.19: 1979321-1979321 on Build GRCh38
rs779221452 Chr.19: 1979310-1979310 on Build GRCh38
rs766073943 Chr.19: 1979281-1979281 on Build GRCh38
rs774985143 Chr.19: 1979226-1979226 on Build GRCh38
rs761260914 Chr.19: 1979255-1979255 on Build GRCh38
rs1043846045 Chr.19: 1979306-1979306 on Build GRCh38
rs11550450 Chr.19: 1979387-1979387 on Build GRCh38
rs758693294 Chr.19: 1979392-1979392 on Build GRCh38
rs374546538 Chr.19: 1979315-1979315 on Build GRCh38
rs368738419 Chr.19: 1979288-1979288 on Build GRCh38
rs768198173 Chr.19: 1979243-1979243 on Build GRCh38
rs138558858 Chr.19: 1979263-1979263 on Build GRCh38
rs896431324 Chr.19: 1979365-1979365 on Build GRCh38
rs764062633 Chr.19: 1979371-1979371 on Build GRCh38
rs778124414 Chr.19: 1979290-1979290 on Build GRCh38
rs757693955 Chr.19: 1979309-1979309 on Build GRCh38
rs757455663 Chr.19: 1979383-1979383 on Build GRCh38
rs759348180 Chr.19: 1979363-1979363 on Build GRCh38
rs771999035 Chr.19: 1979276-1979276 on Build GRCh38

View Full Product Details