Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75304772-75304997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
344219 c.116G>T p.R39I Chr.12: 75304781-75304781 on Build GRCh38
344218 c.812G>T p.R271I Chr.12: 75304781-75304781 on Build GRCh38
6520464 c.21G>T p.R7S Chr.12: 75304876-75304876 on Build GRCh38
943468 c.96G>T p.K32N Chr.12: 75304801-75304801 on Build GRCh38
333908 c.117A>T p.R39S Chr.12: 75304780-75304780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969752546 Chr.12: 75304928-75304928 on Build GRCh38
rs773110136 Chr.12: 75304912-75304912 on Build GRCh38
rs896636178 Chr.12: 75304875-75304875 on Build GRCh38
rs375536734 Chr.12: 75304872-75304872 on Build GRCh38
rs757878927 Chr.12: 75304881-75304881 on Build GRCh38
rs760421258 Chr.12: 75304958-75304958 on Build GRCh38
rs756163297 Chr.12: 75304915-75304915 on Build GRCh38
rs769499853 Chr.12: 75304901-75304901 on Build GRCh38
rs35198123 Chr.12: 75304855-75304855 on Build GRCh38
rs778292034 Chr.12: 75304869-75304869 on Build GRCh38
rs767216438 Chr.12: 75304811-75304811 on Build GRCh38
rs759063481 Chr.12: 75304937-75304937 on Build GRCh38
rs372162631 Chr.12: 75304882-75304882 on Build GRCh38
rs773841222 Chr.12: 75304925-75304925 on Build GRCh38
rs867967333 Chr.12: 75304816-75304816 on Build GRCh38
rs377276215 Chr.12: 75304959-75304959 on Build GRCh38
rs756441621 Chr.12: 75304866-75304866 on Build GRCh38
rs747937077 Chr.12: 75304898-75304898 on Build GRCh38
rs370077776 Chr.12: 75304880-75304880 on Build GRCh38
rs144151792 Chr.12: 75304804-75304804 on Build GRCh38
rs566884715 Chr.12: 75304885-75304885 on Build GRCh38
rs762769547 Chr.12: 75304919-75304919 on Build GRCh38
rs781381737 Chr.12: 75304877-75304877 on Build GRCh38
rs1005057759 Chr.12: 75304815-75304815 on Build GRCh38
rs187500209 Chr.12: 75304921-75304921 on Build GRCh38
rs775857555 Chr.12: 75304886-75304886 on Build GRCh38
rs1001163076 Chr.12: 75304964-75304964 on Build GRCh38
rs752311811 Chr.12: 75304949-75304949 on Build GRCh38
rs778883122 Chr.12: 75304804-75304809 on Build GRCh38
rs746001876 Chr.12: 75304884-75304884 on Build GRCh38
rs767092008 Chr.12: 75304938-75304938 on Build GRCh38

View Full Product Details