Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167135964-167136237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR6
Entrez Gene ID
1235
Gene Name
C-C motif chemokine receptor 6
Gene Aliases
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22
UniGene
Hs.46468
Gene Chromosome Location
Chr.6: 167111807-167139141 on Build GRCh38
Transcripts
NM_004367.5 NM_031409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2854402 c.6C>T p.S2S Chr.6: 167136140-167136140 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9459886 Chr.6: 167136155-167136155 on Build GRCh38
rs761667128 Chr.6: 167136130-167136130 on Build GRCh38
rs922729433 Chr.6: 167136061-167136061 on Build GRCh38
rs555016384 Chr.6: 167136139-167136139 on Build GRCh38
rs776702941 Chr.6: 167136109-167136109 on Build GRCh38
rs772494614 Chr.6: 167136095-167136095 on Build GRCh38
rs754544366 Chr.6: 167136142-167136142 on Build GRCh38
rs993164735 Chr.6: 167135988-167135988 on Build GRCh38
rs368226324 Chr.6: 167136187-167136187 on Build GRCh38
rs778156994 Chr.6: 167136165-167136165 on Build GRCh38
rs998980459 Chr.6: 167136170-167136170 on Build GRCh38
rs765542349 Chr.6: 167136119-167136119 on Build GRCh38
rs3093008 Chr.6: 167136048-167136048 on Build GRCh38
rs911224105 Chr.6: 167135987-167135987 on Build GRCh38
rs878877796 Chr.6: 167136194-167136194 on Build GRCh38
rs201769708 Chr.6: 167136129-167136129 on Build GRCh38
rs775694838 Chr.6: 167136078-167136078 on Build GRCh38
rs754589241 Chr.6: 167136040-167136044 on Build GRCh38
rs745938330 Chr.6: 167136195-167136195 on Build GRCh38
rs17860849 Chr.6: 167136062-167136062 on Build GRCh38
rs1014068342 Chr.6: 167136197-167136197 on Build GRCh38
rs112836572 Chr.6: 167136169-167136169 on Build GRCh38
rs200741825 Chr.6: 167136164-167136164 on Build GRCh38
rs747335022 Chr.6: 167136171-167136171 on Build GRCh38
rs775830961 Chr.6: 167136206-167136206 on Build GRCh38
rs753362182 Chr.6: 167136065-167136065 on Build GRCh38
rs888584548 Chr.6: 167136100-167136100 on Build GRCh38
rs372109980 Chr.6: 167136205-167136205 on Build GRCh38
rs759697824 Chr.6: 167136110-167136110 on Build GRCh38
rs771369954 Chr.6: 167136106-167136106 on Build GRCh38
rs1013966200 Chr.6: 167136145-167136145 on Build GRCh38
rs3093009 Chr.6: 167135989-167135989 on Build GRCh38
rs189192805 Chr.6: 167136001-167136001 on Build GRCh38
rs747841027 Chr.6: 167136156-167136156 on Build GRCh38
rs766231476 Chr.6: 167136141-167136141 on Build GRCh38
rs970147769 Chr.6: 167136010-167136010 on Build GRCh38
rs762913226 Chr.6: 167136140-167136140 on Build GRCh38
rs761191877 Chr.6: 167136102-167136102 on Build GRCh38
rs998971859 Chr.6: 167136168-167136168 on Build GRCh38
rs193057414 Chr.6: 167136172-167136172 on Build GRCh38
rs773564155 Chr.6: 167136097-167136097 on Build GRCh38

View Full Product Details