Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33677090-33677343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545485 c.1036C>T p.P346S Chr.20: 33677135-33677135 on Build GRCh38
3545486 c.880G>A p.D294N Chr.20: 33677291-33677291 on Build GRCh38
2757812 c.1004C>A p.P335Q Chr.20: 33677167-33677167 on Build GRCh38
6092803 c.916A>G p.I306V Chr.20: 33677255-33677255 on Build GRCh38
7079931 c.1053C>G p.L351L Chr.20: 33677118-33677118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747031927 Chr.20: 33677161-33677163 on Build GRCh38
rs535573433 Chr.20: 33677245-33677245 on Build GRCh38
rs752029737 Chr.20: 33677156-33677156 on Build GRCh38
rs756501085 Chr.20: 33677313-33677313 on Build GRCh38
rs774185605 Chr.20: 33677136-33677136 on Build GRCh38
rs764704208 Chr.20: 33677283-33677283 on Build GRCh38
rs267605892 Chr.20: 33677292-33677292 on Build GRCh38
rs145759508 Chr.20: 33677135-33677135 on Build GRCh38
rs757620294 Chr.20: 33677159-33677159 on Build GRCh38
rs776058128 Chr.20: 33677208-33677210 on Build GRCh38
rs759041493 Chr.20: 33677280-33677280 on Build GRCh38
rs775614866 Chr.20: 33677118-33677118 on Build GRCh38
rs773197012 Chr.20: 33677249-33677249 on Build GRCh38
rs762296081 Chr.20: 33677291-33677291 on Build GRCh38
rs770643353 Chr.20: 33677264-33677264 on Build GRCh38
rs778884599 Chr.20: 33677236-33677236 on Build GRCh38
rs1031106445 Chr.20: 33677302-33677302 on Build GRCh38
rs752075116 Chr.20: 33677289-33677289 on Build GRCh38
rs771807428 Chr.20: 33677140-33677140 on Build GRCh38
rs764626563 Chr.20: 33677155-33677155 on Build GRCh38
rs770856342 Chr.20: 33677166-33677168 on Build GRCh38
rs759073647 Chr.20: 33677238-33677264 on Build GRCh38
rs753343055 Chr.20: 33677151-33677151 on Build GRCh38
rs111268294 Chr.20: 33677160-33677160 on Build GRCh38
rs765976516 Chr.20: 33677144-33677144 on Build GRCh38
rs575839838 Chr.20: 33677295-33677295 on Build GRCh38
rs553413048 Chr.20: 33677201-33677201 on Build GRCh38
rs554294623 Chr.20: 33677265-33677265 on Build GRCh38
rs749348639 Chr.20: 33677218-33677218 on Build GRCh38
rs750641087 Chr.20: 33677170-33677170 on Build GRCh38
rs768640915 Chr.20: 33677234-33677234 on Build GRCh38
rs144481343 Chr.20: 33677203-33677203 on Build GRCh38
rs750718703 Chr.20: 33677307-33677307 on Build GRCh38
rs138360482 Chr.20: 33677154-33677154 on Build GRCh38
rs149272498 Chr.20: 33677168-33677168 on Build GRCh38
rs944228695 Chr.20: 33677238-33677238 on Build GRCh38
rs3213174 Chr.20: 33677239-33677239 on Build GRCh38
rs867122904 Chr.20: 33677296-33677296 on Build GRCh38
rs760562883 Chr.20: 33677250-33677250 on Build GRCh38
rs373179329 Chr.20: 33677142-33677142 on Build GRCh38
rs761616906 Chr.20: 33677137-33677137 on Build GRCh38
rs574956843 Chr.20: 33677278-33677278 on Build GRCh38
rs993029218 Chr.20: 33677129-33677129 on Build GRCh38
rs749340138 Chr.20: 33677123-33677123 on Build GRCh38
rs564825532 Chr.20: 33677143-33677143 on Build GRCh38

View Full Product Details