Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155239937-155240164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359123 c.226T>C p.F76L Chr.1: 155239967-155239967 on Build GRCh38
7111909 c.187G>C p.D63H Chr.1: 155240006-155240006 on Build GRCh38
4022968 c.155C>T p.S52L Chr.1: 155240038-155240038 on Build GRCh38
161214 c.204G>A p.P68P Chr.1: 155239989-155239989 on Build GRCh38
6870043 c.211C>T p.P71S Chr.1: 155239982-155239982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145888253 Chr.1: 155240008-155240008 on Build GRCh38
rs760804175 Chr.1: 155240115-155240115 on Build GRCh38
rs749118277 Chr.1: 155240087-155240087 on Build GRCh38
rs151093421 Chr.1: 155239977-155239977 on Build GRCh38
rs368786234 Chr.1: 155239962-155239962 on Build GRCh38
rs757834551 Chr.1: 155239987-155239987 on Build GRCh38
rs752857428 Chr.1: 155239960-155239960 on Build GRCh38
rs779377390 Chr.1: 155240069-155240069 on Build GRCh38
rs750891392 Chr.1: 155240061-155240061 on Build GRCh38
rs767693527 Chr.1: 155239956-155239956 on Build GRCh38
rs878853318 Chr.1: 155240026-155240026 on Build GRCh38
rs747971975 Chr.1: 155240020-155240020 on Build GRCh38
rs121908302 Chr.1: 155240033-155240033 on Build GRCh38
rs779258874 Chr.1: 155239970-155239970 on Build GRCh38
rs750712570 Chr.1: 155239989-155239989 on Build GRCh38
rs75954905 Chr.1: 155239965-155239965 on Build GRCh38
rs763010874 Chr.1: 155240031-155240031 on Build GRCh38
rs146774384 Chr.1: 155239961-155239961 on Build GRCh38
rs770922837 Chr.1: 155240095-155240095 on Build GRCh38
rs773007510 Chr.1: 155240029-155240029 on Build GRCh38
rs768903489 Chr.1: 155240121-155240121 on Build GRCh38
rs769900428 Chr.1: 155239974-155239974 on Build GRCh38
rs750990483 Chr.1: 155239978-155239978 on Build GRCh38
rs371592589 Chr.1: 155239972-155239972 on Build GRCh38
rs199565854 Chr.1: 155240122-155240122 on Build GRCh38
rs142761046 Chr.1: 155240057-155240057 on Build GRCh38
rs190207858 Chr.1: 155240072-155240072 on Build GRCh38
rs200378040 Chr.1: 155240077-155240077 on Build GRCh38
rs758949381 Chr.1: 155240067-155240067 on Build GRCh38
rs775652188 Chr.1: 155240045-155240045 on Build GRCh38
rs145773486 Chr.1: 155240025-155240025 on Build GRCh38
rs747853210 Chr.1: 155240076-155240076 on Build GRCh38
rs543005460 Chr.1: 155240053-155240053 on Build GRCh38
rs760930573 Chr.1: 155240048-155240048 on Build GRCh38
rs141061530 Chr.1: 155239990-155239990 on Build GRCh38
rs765693058 Chr.1: 155240060-155240060 on Build GRCh38
rs761621516 Chr.1: 155239969-155239971 on Build GRCh38
rs777624686 Chr.1: 155240078-155240078 on Build GRCh38
rs373363286 Chr.1: 155239969-155239969 on Build GRCh38
rs751095441 Chr.1: 155240071-155240071 on Build GRCh38
rs185884197 Chr.1: 155240049-155240049 on Build GRCh38
rs74953658 Chr.1: 155240004-155240004 on Build GRCh38
rs756264143 Chr.1: 155240037-155240037 on Build GRCh38

View Full Product Details