Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41621916-41622175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
445056 c.31G>C p.E11Q Chr.22: 41622035-41622035 on Build GRCh38
6824148 c.59_61delAAG p.E22delE Chr.22: 41622063-41622065 on Build GRCh38
1232992 c.2T>C p.M1T Chr.22: 41622006-41622006 on Build GRCh38
5013695 c.82+1G>A p.? Chr.22: 41622087-41622087 on Build GRCh38
726992 c.60A>G p.Q20Q Chr.22: 41622064-41622064 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751053053 Chr.22: 41622138-41622138 on Build GRCh38
rs764558937 Chr.22: 41622119-41622119 on Build GRCh38
rs760110269 Chr.22: 41621980-41621980 on Build GRCh38
rs759573926 Chr.22: 41622105-41622105 on Build GRCh38
rs752218326 Chr.22: 41622124-41622125 on Build GRCh38
rs368257845 Chr.22: 41621990-41621990 on Build GRCh38
rs757781595 Chr.22: 41622003-41622003 on Build GRCh38
rs761064095 Chr.22: 41622116-41622116 on Build GRCh38
rs372416521 Chr.22: 41622034-41622034 on Build GRCh38
rs758094570 Chr.22: 41622131-41622131 on Build GRCh38
rs778696905 Chr.22: 41621991-41621991 on Build GRCh38
rs561790680 Chr.22: 41621939-41621939 on Build GRCh38
rs766481862 Chr.22: 41621965-41621965 on Build GRCh38
rs764700410 Chr.22: 41622124-41622128 on Build GRCh38
rs750004711 Chr.22: 41622130-41622130 on Build GRCh38
rs369186241 Chr.22: 41622084-41622084 on Build GRCh38
rs933286005 Chr.22: 41622114-41622114 on Build GRCh38
rs528964413 Chr.22: 41621964-41621964 on Build GRCh38
rs753314269 Chr.22: 41622115-41622115 on Build GRCh38
rs770865015 Chr.22: 41622102-41622102 on Build GRCh38
rs763264491 Chr.22: 41622090-41622090 on Build GRCh38
rs753079862 Chr.22: 41621983-41621983 on Build GRCh38
rs912566256 Chr.22: 41622145-41622145 on Build GRCh38
rs200736808 Chr.22: 41622024-41622024 on Build GRCh38
rs774466382 Chr.22: 41622104-41622104 on Build GRCh38
rs768562473 Chr.22: 41622043-41622043 on Build GRCh38
rs376715887 Chr.22: 41622040-41622040 on Build GRCh38
rs773477333 Chr.22: 41621960-41621960 on Build GRCh38
rs750105679 Chr.22: 41622002-41622002 on Build GRCh38
rs971945322 Chr.22: 41622100-41622100 on Build GRCh38
rs547529160 Chr.22: 41622057-41622057 on Build GRCh38
rs747920624 Chr.22: 41622067-41622067 on Build GRCh38
rs765549166 Chr.22: 41621958-41621958 on Build GRCh38
rs375802511 Chr.22: 41622134-41622134 on Build GRCh38
rs761649266 Chr.22: 41621959-41621959 on Build GRCh38
rs769585677 Chr.22: 41622070-41622070 on Build GRCh38
rs751968379 Chr.22: 41621966-41621966 on Build GRCh38
rs373933575 Chr.22: 41622108-41622108 on Build GRCh38
rs768186462 Chr.22: 41621981-41621981 on Build GRCh38

View Full Product Details