Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75222741-75223014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRB1
Entrez Gene ID
1504
Gene Name
chymotrypsinogen B1
Gene Aliases
CTRB
UniGene
Hs.610926
Gene Chromosome Location
Chr.16: 75218986-75224924 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6324921 c.66C>T p.P22P Chr.16: 75222781-75222781 on Build GRCh38
5420248 c.53-6C>A p.? Chr.16: 75222762-75222762 on Build GRCh38
6350909 c.102C>T p.I34I Chr.16: 75222817-75222817 on Build GRCh38
3957826 c.140G>T p.W47L Chr.16: 75222855-75222855 on Build GRCh38
5692590 c.109G>A p.G37R Chr.16: 75222824-75222824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371359313 Chr.16: 75222977-75222977 on Build GRCh38
rs1018640521 Chr.16: 75222855-75222855 on Build GRCh38
rs965544442 Chr.16: 75222860-75222860 on Build GRCh38
rs1004348093 Chr.16: 75222762-75222762 on Build GRCh38
rs753039239 Chr.16: 75222877-75222877 on Build GRCh38
rs775169798 Chr.16: 75222821-75222821 on Build GRCh38
rs777302486 Chr.16: 75222794-75222794 on Build GRCh38
rs917121804 Chr.16: 75222793-75222793 on Build GRCh38
rs760725877 Chr.16: 75222837-75222837 on Build GRCh38
rs765593873 Chr.16: 75222785-75222785 on Build GRCh38
rs979177970 Chr.16: 75222962-75222962 on Build GRCh38
rs765540919 Chr.16: 75222865-75222865 on Build GRCh38
rs779868260 Chr.16: 75222971-75222971 on Build GRCh38
rs747955269 Chr.16: 75222981-75222981 on Build GRCh38
rs200004817 Chr.16: 75222760-75222761 on Build GRCh38
rs576908589 Chr.16: 75222765-75222765 on Build GRCh38
rs372989485 Chr.16: 75222775-75222775 on Build GRCh38
rs562578052 Chr.16: 75222818-75222818 on Build GRCh38
rs772224212 Chr.16: 75222835-75222835 on Build GRCh38
rs533494856 Chr.16: 75222978-75222978 on Build GRCh38
rs745750472 Chr.16: 75222943-75222943 on Build GRCh38
rs535281828 Chr.16: 75222763-75222763 on Build GRCh38
rs954292179 Chr.16: 75222898-75222898 on Build GRCh38
rs773252714 Chr.16: 75222993-75222993 on Build GRCh38
rs201073029 Chr.16: 75222950-75222950 on Build GRCh38
rs372716369 Chr.16: 75222803-75222803 on Build GRCh38
rs756759171 Chr.16: 75222916-75222916 on Build GRCh38
rs199938071 Chr.16: 75222846-75222846 on Build GRCh38
rs746564301 Chr.16: 75222796-75222796 on Build GRCh38
rs1029011165 Chr.16: 75222891-75222891 on Build GRCh38
rs199776091 Chr.16: 75222817-75222817 on Build GRCh38
rs748380432 Chr.16: 75222833-75222833 on Build GRCh38
rs766463599 Chr.16: 75222841-75222841 on Build GRCh38
rs575650101 Chr.16: 75222836-75222836 on Build GRCh38
rs201853088 Chr.16: 75222927-75222927 on Build GRCh38
rs550027858 Chr.16: 75222992-75222992 on Build GRCh38
rs752358929 Chr.16: 75222787-75222787 on Build GRCh38
rs757245079 Chr.16: 75222802-75222802 on Build GRCh38
rs527574397 Chr.16: 75222892-75222892 on Build GRCh38
rs760071006 Chr.16: 75222863-75222863 on Build GRCh38
rs751235075 Chr.16: 75222897-75222897 on Build GRCh38
rs561650096 Chr.16: 75222886-75222886 on Build GRCh38
rs369201413 Chr.16: 75222788-75222788 on Build GRCh38
rs369595328 Chr.16: 75222764-75222764 on Build GRCh38
rs769722495 Chr.16: 75222760-75222760 on Build GRCh38

View Full Product Details