Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30710309-30710550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019864.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2205129 c.2903G>A p.R968H Chr.12: 30710383-30710383 on Build GRCh38
7277097 c.2878A>G p.R960G Chr.12: 30710449-30710449 on Build GRCh38
6620007 c.2905A>G p.S969G Chr.12: 30710381-30710381 on Build GRCh38
5696414 c.2955G>T p.L985L Chr.12: 30710331-30710331 on Build GRCh38
6620001 c.2959G>A p.V987I Chr.12: 30710327-30710327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747195058 Chr.12: 30710498-30710498 on Build GRCh38
rs201095477 Chr.12: 30710412-30710412 on Build GRCh38
rs372208056 Chr.12: 30710337-30710337 on Build GRCh38
rs79552778 Chr.12: 30710379-30710379 on Build GRCh38
rs761381989 Chr.12: 30710432-30710432 on Build GRCh38
rs973720071 Chr.12: 30710470-30710470 on Build GRCh38
rs965803693 Chr.12: 30710331-30710331 on Build GRCh38
rs767335762 Chr.12: 30710433-30710433 on Build GRCh38
rs756585609 Chr.12: 30710446-30710446 on Build GRCh38
rs927003563 Chr.12: 30710409-30710409 on Build GRCh38
rs140741615 Chr.12: 30710336-30710336 on Build GRCh38
rs772253137 Chr.12: 30710383-30710383 on Build GRCh38
rs375234262 Chr.12: 30710361-30710361 on Build GRCh38
rs371328465 Chr.12: 30710424-30710424 on Build GRCh38
rs750076759 Chr.12: 30710445-30710445 on Build GRCh38
rs878879989 Chr.12: 30710357-30710357 on Build GRCh38
rs535610393 Chr.12: 30710346-30710346 on Build GRCh38
rs759274017 Chr.12: 30710393-30710393 on Build GRCh38
rs775209566 Chr.12: 30710499-30710499 on Build GRCh38
rs755379630 Chr.12: 30710455-30710455 on Build GRCh38
rs780659467 Chr.12: 30710340-30710340 on Build GRCh38
rs367800608 Chr.12: 30710384-30710384 on Build GRCh38
rs762953715 Chr.12: 30710520-30710520 on Build GRCh38
rs200624700 Chr.12: 30710486-30710486 on Build GRCh38
rs754416397 Chr.12: 30710451-30710451 on Build GRCh38
rs775373364 Chr.12: 30710411-30710411 on Build GRCh38
rs761265039 Chr.12: 30710513-30710513 on Build GRCh38
rs769605672 Chr.12: 30710406-30710406 on Build GRCh38
rs976348698 Chr.12: 30710347-30710347 on Build GRCh38
rs763676110 Chr.12: 30710423-30710423 on Build GRCh38
rs1044833 Chr.12: 30710471-30710471 on Build GRCh38
rs772173740 Chr.12: 30710490-30710490 on Build GRCh38
rs779052048 Chr.12: 30710472-30710472 on Build GRCh38
rs780524389 Chr.12: 30710447-30710447 on Build GRCh38
rs778912498 Chr.12: 30710378-30710378 on Build GRCh38
rs778012995 Chr.12: 30710497-30710497 on Build GRCh38

View Full Product Details