Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159781284-159781539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP23
Entrez Gene ID
54935
Gene Name
dual specificity phosphatase 23
Gene Aliases
DUSP25, LDP-3, MOSP, VHZ
UniGene
Hs.425801
Gene Chromosome Location
Chr.1: 159780946-159782546 on Build GRCh38
Transcripts
NM_001319658.1 NM_001319659.1 NM_017823.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409141 c.208G>C p.A70P Chr.1: 159781308-159781308 on Build GRCh38
6368671 c.188T>C p.I63T Chr.1: 159781288-159781288 on Build GRCh38
1601091 c.241G>T p.V81L Chr.1: 159781341-159781341 on Build GRCh38
6431003 c.198C>A p.F66L Chr.1: 159781298-159781298 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959484106 Chr.1: 159781331-159781331 on Build GRCh38
rs981446053 Chr.1: 159781359-159781359 on Build GRCh38
rs1001201071 Chr.1: 159781311-159781311 on Build GRCh38
rs758361500 Chr.1: 159781333-159781333 on Build GRCh38
rs1036619330 Chr.1: 159781498-159781498 on Build GRCh38
rs571901654 Chr.1: 159781358-159781358 on Build GRCh38
rs148687167 Chr.1: 159781314-159781314 on Build GRCh38
rs908790974 Chr.1: 159781360-159781360 on Build GRCh38
rs747774801 Chr.1: 159781388-159781388 on Build GRCh38
rs902183100 Chr.1: 159781512-159781512 on Build GRCh38
rs974748902 Chr.1: 159781386-159781386 on Build GRCh38
rs1012971765 Chr.1: 159781340-159781340 on Build GRCh38
rs931670186 Chr.1: 159781449-159781452 on Build GRCh38
rs751586016 Chr.1: 159781366-159781366 on Build GRCh38
rs943679492 Chr.1: 159781420-159781420 on Build GRCh38
rs1023033264 Chr.1: 159781347-159781347 on Build GRCh38
rs912355796 Chr.1: 159781415-159781415 on Build GRCh38
rs1054124137 Chr.1: 159781518-159781518 on Build GRCh38
rs780115677 Chr.1: 159781354-159781354 on Build GRCh38
rs932842383 Chr.1: 159781392-159781392 on Build GRCh38
rs920078551 Chr.1: 159781390-159781390 on Build GRCh38
rs922919885 Chr.1: 159781443-159781443 on Build GRCh38
rs1050172809 Chr.1: 159781453-159781453 on Build GRCh38
rs1042043430 Chr.1: 159781480-159781480 on Build GRCh38
rs564314890 Chr.1: 159781327-159781327 on Build GRCh38
rs200809170 Chr.1: 159781319-159781319 on Build GRCh38
rs564341682 Chr.1: 159781462-159781462 on Build GRCh38
rs549857570 Chr.1: 159781516-159781516 on Build GRCh38
rs142235884 Chr.1: 159781374-159781374 on Build GRCh38
rs769332056 Chr.1: 159781405-159781405 on Build GRCh38
rs941842179 Chr.1: 159781490-159781490 on Build GRCh38
rs977026448 Chr.1: 159781416-159781416 on Build GRCh38
rs936575778 Chr.1: 159781515-159781515 on Build GRCh38
rs780622883 Chr.1: 159781377-159781377 on Build GRCh38

View Full Product Details