Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 21909307-21909570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK2
Entrez Gene ID
9046
Gene Name
docking protein 2
Gene Aliases
p56DOK, p56dok-2
UniGene
Hs.71215
Gene Chromosome Location
Chr.8: 21908873-21913717 on Build GRCh38
Transcripts
NM_001317800.1 NM_003974.3 NM_201349.2 XM_005273680.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5655594 c.1070A>C p.Y357S Chr.8: 21909480-21909480 on Build GRCh38
6495205 c.1137delT p.G380fs*28 Chr.8: 21909413-21909413 on Build GRCh38
6495204 c.1180T>G p.S394A Chr.8: 21909370-21909370 on Build GRCh38
1699782 c.1079C>T p.P360L Chr.8: 21909471-21909471 on Build GRCh38
1755662 c.1189C>G p.Q397E Chr.8: 21909361-21909361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749737261 Chr.8: 21909477-21909477 on Build GRCh38
rs781263286 Chr.8: 21909361-21909361 on Build GRCh38
rs757494381 Chr.8: 21909532-21909532 on Build GRCh38
rs760777734 Chr.8: 21909530-21909530 on Build GRCh38
rs1052886594 Chr.8: 21909462-21909462 on Build GRCh38
rs139826199 Chr.8: 21909510-21909510 on Build GRCh38
rs759637576 Chr.8: 21909433-21909444 on Build GRCh38
rs912748742 Chr.8: 21909419-21909419 on Build GRCh38
rs775153682 Chr.8: 21909385-21909385 on Build GRCh38
rs779421891 Chr.8: 21909534-21909534 on Build GRCh38
rs775382276 Chr.8: 21909437-21909437 on Build GRCh38
rs768030217 Chr.8: 21909459-21909459 on Build GRCh38
rs7824139 Chr.8: 21909544-21909544 on Build GRCh38
rs752753850 Chr.8: 21909473-21909473 on Build GRCh38
rs146656524 Chr.8: 21909449-21909449 on Build GRCh38
rs143272640 Chr.8: 21909435-21909435 on Build GRCh38
rs776012346 Chr.8: 21909495-21909495 on Build GRCh38
rs150641697 Chr.8: 21909505-21909505 on Build GRCh38
rs765244197 Chr.8: 21909497-21909498 on Build GRCh38
rs140097138 Chr.8: 21909470-21909470 on Build GRCh38
rs2242241 Chr.8: 21909370-21909370 on Build GRCh38
rs764966266 Chr.8: 21909401-21909401 on Build GRCh38
rs766251181 Chr.8: 21909413-21909413 on Build GRCh38
rs200583668 Chr.8: 21909382-21909382 on Build GRCh38
rs762782810 Chr.8: 21909513-21909513 on Build GRCh38
rs780142730 Chr.8: 21909343-21909343 on Build GRCh38
rs746558832 Chr.8: 21909458-21909458 on Build GRCh38
rs201556605 Chr.8: 21909354-21909354 on Build GRCh38
rs758848269 Chr.8: 21909538-21909538 on Build GRCh38
rs17853066 Chr.8: 21909389-21909389 on Build GRCh38
rs369392176 Chr.8: 21909388-21909388 on Build GRCh38
rs751523081 Chr.8: 21909506-21909506 on Build GRCh38
rs747640366 Chr.8: 21909502-21909502 on Build GRCh38
rs771624166 Chr.8: 21909445-21909445 on Build GRCh38
rs200038363 Chr.8: 21909523-21909523 on Build GRCh38
rs773008623 Chr.8: 21909461-21909461 on Build GRCh38
rs756318240 Chr.8: 21909433-21909433 on Build GRCh38
rs773574938 Chr.8: 21909372-21909372 on Build GRCh38
rs772815734 Chr.8: 21909512-21909512 on Build GRCh38
rs755017117 Chr.8: 21909418-21909418 on Build GRCh38
rs375220071 Chr.8: 21909469-21909469 on Build GRCh38
rs1045661807 Chr.8: 21909480-21909480 on Build GRCh38
rs200171615 Chr.8: 21909408-21909408 on Build GRCh38
rs565835482 Chr.8: 21909424-21909424 on Build GRCh38
rs761719114 Chr.8: 21909396-21909396 on Build GRCh38
rs149795299 Chr.8: 21909515-21909515 on Build GRCh38
rs372534431 Chr.8: 21909531-21909531 on Build GRCh38
rs143948915 Chr.8: 21909476-21909476 on Build GRCh38
rs779613815 Chr.8: 21909478-21909478 on Build GRCh38
rs536644394 Chr.8: 21909490-21909490 on Build GRCh38
rs772594193 Chr.8: 21909492-21909492 on Build GRCh38
rs754858807 Chr.8: 21909471-21909471 on Build GRCh38
rs200790064 Chr.8: 21909516-21909516 on Build GRCh38
rs145725971 Chr.8: 21909522-21909522 on Build GRCh38
rs758287012 Chr.8: 21909340-21909340 on Build GRCh38
rs377248888 Chr.8: 21909537-21909537 on Build GRCh38
rs766181483 Chr.8: 21909514-21909514 on Build GRCh38
rs763583482 Chr.8: 21909374-21909374 on Build GRCh38
rs757140092 Chr.8: 21909544-21909544 on Build GRCh38
rs768920141 Chr.8: 21909358-21909358 on Build GRCh38
rs751700999 Chr.8: 21909417-21909417 on Build GRCh38
rs202013016 Chr.8: 21909460-21909460 on Build GRCh38
rs962546682 Chr.8: 21909539-21909539 on Build GRCh38
rs748582902 Chr.8: 21909369-21909369 on Build GRCh38
rs61757360 Chr.8: 21909335-21909335 on Build GRCh38

View Full Product Details