Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15239519-15239780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3529580 c.3445C>T p.R1149W Chr.19: 15239659-15239659 on Build GRCh38
4639438 c.3430G>A p.V1144I Chr.19: 15239674-15239674 on Build GRCh38
6912332 c.3337_3339delGAG p.E1113delE Chr.19: 15239765-15239767 on Build GRCh38
6612717 c.3444G>T p.Q1148H Chr.19: 15239660-15239660 on Build GRCh38
3283220 c.3449C>T p.P1150L Chr.19: 15239519-15239519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59758739 Chr.19: 15239540-15239540 on Build GRCh38
rs755212384 Chr.19: 15239610-15239610 on Build GRCh38
rs750248387 Chr.19: 15239585-15239585 on Build GRCh38
rs976668161 Chr.19: 15239723-15239723 on Build GRCh38
rs201671277 Chr.19: 15239680-15239680 on Build GRCh38
rs776104741 Chr.19: 15239569-15239569 on Build GRCh38
rs1055671896 Chr.19: 15239623-15239623 on Build GRCh38
rs567564484 Chr.19: 15239601-15239601 on Build GRCh38
rs749421543 Chr.19: 15239650-15239650 on Build GRCh38
rs376743001 Chr.19: 15239706-15239706 on Build GRCh38
rs780262620 Chr.19: 15239649-15239649 on Build GRCh38
rs200957651 Chr.19: 15239674-15239674 on Build GRCh38
rs772188667 Chr.19: 15239642-15239643 on Build GRCh38
rs746826210 Chr.19: 15239554-15239554 on Build GRCh38
rs748633137 Chr.19: 15239721-15239721 on Build GRCh38
rs761598463 Chr.19: 15239693-15239693 on Build GRCh38
rs200345800 Chr.19: 15239617-15239617 on Build GRCh38
rs376162076 Chr.19: 15239716-15239716 on Build GRCh38
rs1047396764 Chr.19: 15239671-15239671 on Build GRCh38
rs201935715 Chr.19: 15239733-15239733 on Build GRCh38
rs946602520 Chr.19: 15239552-15239552 on Build GRCh38
rs553993519 Chr.19: 15239700-15239700 on Build GRCh38
rs201578775 Chr.19: 15239675-15239675 on Build GRCh38
rs201917407 Chr.19: 15239561-15239561 on Build GRCh38
rs757468381 Chr.19: 15239555-15239555 on Build GRCh38
rs556458475 Chr.19: 15239627-15239627 on Build GRCh38
rs967545322 Chr.19: 15239682-15239682 on Build GRCh38
rs199517872 Chr.19: 15239626-15239626 on Build GRCh38
rs757054890 Chr.19: 15239640-15239640 on Build GRCh38
rs570161414 Chr.19: 15239631-15239631 on Build GRCh38
rs1040505052 Chr.19: 15239551-15239551 on Build GRCh38
rs772431443 Chr.19: 15239737-15239737 on Build GRCh38
rs762462881 Chr.19: 15239758-15239758 on Build GRCh38
rs762435043 Chr.19: 15239684-15239684 on Build GRCh38
rs199544374 Chr.19: 15239678-15239678 on Build GRCh38
rs777644859 Chr.19: 15239543-15239543 on Build GRCh38
rs60057301 Chr.19: 15239556-15239556 on Build GRCh38
rs200703650 Chr.19: 15239715-15239715 on Build GRCh38
rs200095218 Chr.19: 15239711-15239711 on Build GRCh38
rs377217681 Chr.19: 15239644-15239644 on Build GRCh38
rs370451418 Chr.19: 15239652-15239652 on Build GRCh38
rs547584115 Chr.19: 15239591-15239591 on Build GRCh38
rs747411770 Chr.19: 15239747-15239747 on Build GRCh38
rs200850373 Chr.19: 15239638-15239638 on Build GRCh38
rs762152914 Chr.19: 15239615-15239615 on Build GRCh38
rs1032745641 Chr.19: 15239690-15239690 on Build GRCh38
rs376346739 Chr.19: 15239572-15239572 on Build GRCh38
rs746128795 Chr.19: 15239647-15239647 on Build GRCh38
rs540369634 Chr.19: 15239694-15239694 on Build GRCh38
rs944905000 Chr.19: 15239546-15239546 on Build GRCh38
rs573909313 Chr.19: 15239738-15239738 on Build GRCh38
rs759208540 Chr.19: 15239676-15239676 on Build GRCh38
rs765914805 Chr.19: 15239562-15239562 on Build GRCh38
rs778802854 Chr.19: 15239549-15239573 on Build GRCh38
rs779348570 Chr.19: 15239659-15239659 on Build GRCh38
rs200628536 Chr.19: 15239679-15239679 on Build GRCh38
rs199839337 Chr.19: 15239743-15239743 on Build GRCh38
rs747867996 Chr.19: 15239617-15239618 on Build GRCh38
rs764075971 Chr.19: 15239685-15239685 on Build GRCh38
rs775177082 Chr.19: 15239757-15239757 on Build GRCh38
rs368982771 Chr.19: 15239618-15239618 on Build GRCh38
rs779410862 Chr.19: 15239720-15239720 on Build GRCh38
rs199735840 Chr.19: 15239624-15239624 on Build GRCh38
rs751462158 Chr.19: 15239686-15239686 on Build GRCh38
rs774749936 Chr.19: 15239611-15239611 on Build GRCh38
rs756217610 Chr.19: 15239648-15239648 on Build GRCh38

View Full Product Details