Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75284796-75285057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020040.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575738 c.1487A>G p.N496S Chr.12: 75285046-75285046 on Build GRCh38
7359474 c.1513G>T p.E505* Chr.12: 75285020-75285020 on Build GRCh38
6138076 c.1495G>T p.G499C Chr.12: 75285038-75285038 on Build GRCh38
4528655 c.1554G>A p.R518R Chr.12: 75284979-75284979 on Build GRCh38
4973742 c.1567C>T p.R523* Chr.12: 75284966-75284966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764447136 Chr.12: 75285011-75285011 on Build GRCh38
rs749900141 Chr.12: 75285010-75285010 on Build GRCh38
rs112509075 Chr.12: 75284915-75284915 on Build GRCh38
rs757338387 Chr.12: 75284935-75284935 on Build GRCh38
rs1008592285 Chr.12: 75284848-75284848 on Build GRCh38
rs780831319 Chr.12: 75284949-75284949 on Build GRCh38
rs754721898 Chr.12: 75284948-75284948 on Build GRCh38
rs143527877 Chr.12: 75284988-75284988 on Build GRCh38
rs147185184 Chr.12: 75285005-75285005 on Build GRCh38
rs1056667918 Chr.12: 75284975-75284975 on Build GRCh38
rs900123722 Chr.12: 75284906-75284906 on Build GRCh38
rs529829437 Chr.12: 75284864-75284864 on Build GRCh38
rs748750102 Chr.12: 75284967-75284967 on Build GRCh38
rs770596009 Chr.12: 75284972-75284972 on Build GRCh38
rs777123293 Chr.12: 75285036-75285036 on Build GRCh38
rs368171441 Chr.12: 75284973-75284973 on Build GRCh38
rs948834616 Chr.12: 75284860-75284860 on Build GRCh38
rs113382753 Chr.12: 75284922-75284922 on Build GRCh38
rs748016719 Chr.12: 75284958-75284958 on Build GRCh38
rs757183829 Chr.12: 75285000-75285002 on Build GRCh38
rs749224602 Chr.12: 75284924-75284924 on Build GRCh38
rs761156944 Chr.12: 75285008-75285008 on Build GRCh38
rs377208231 Chr.12: 75284828-75284828 on Build GRCh38
rs779045003 Chr.12: 75284936-75284936 on Build GRCh38
rs137966754 Chr.12: 75284961-75284961 on Build GRCh38
rs1052535388 Chr.12: 75284823-75284823 on Build GRCh38
rs764725871 Chr.12: 75284919-75284919 on Build GRCh38
rs201652520 Chr.12: 75284966-75284966 on Build GRCh38
rs766712873 Chr.12: 75284960-75284960 on Build GRCh38
rs750679806 Chr.12: 75284942-75284942 on Build GRCh38
rs560359375 Chr.12: 75284929-75284929 on Build GRCh38
rs909187893 Chr.12: 75284852-75284852 on Build GRCh38
rs960789862 Chr.12: 75284986-75284986 on Build GRCh38
rs891584476 Chr.12: 75284832-75284832 on Build GRCh38
rs759239804 Chr.12: 75284980-75284980 on Build GRCh38
rs145711273 Chr.12: 75284863-75284863 on Build GRCh38

View Full Product Details