Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65040721-65040992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3

Gene Symbol
MAX
Entrez Gene ID
4149
Gene Name
MYC associated factor X
Gene Aliases
bHLHd4
UniGene
Hs.285354
Gene Chromosome Location
Chr.14: 65006101-65102695 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402327 c.721G>T p.G241W Chr.14: 65040818-65040818 on Build GRCh38
4051904 c.772G>A p.A258T Chr.14: 65040869-65040869 on Build GRCh38
3386639 c.810G>A p.L270L Chr.14: 65040907-65040907 on Build GRCh38
6498785 c.732G>T p.G244G Chr.14: 65040829-65040829 on Build GRCh38
4051901 c.701A>G p.N234S Chr.14: 65040798-65040798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757444379 Chr.14: 65040957-65040957 on Build GRCh38
rs755510047 Chr.14: 65040781-65040781 on Build GRCh38
rs776308370 Chr.14: 65040875-65040875 on Build GRCh38
rs955568247 Chr.14: 65040951-65040951 on Build GRCh38
rs369112788 Chr.14: 65040782-65040782 on Build GRCh38
rs752063880 Chr.14: 65040776-65040776 on Build GRCh38
rs915456962 Chr.14: 65040878-65040878 on Build GRCh38
rs530614536 Chr.14: 65040883-65040883 on Build GRCh38
rs757798314 Chr.14: 65040856-65040856 on Build GRCh38
rs781068835 Chr.14: 65040863-65040863 on Build GRCh38
rs754135429 Chr.14: 65040954-65040954 on Build GRCh38
rs45563031 Chr.14: 65040936-65040936 on Build GRCh38
rs767024921 Chr.14: 65040768-65040768 on Build GRCh38
rs764240926 Chr.14: 65040950-65040950 on Build GRCh38
rs376359664 Chr.14: 65040904-65040904 on Build GRCh38
rs761322343 Chr.14: 65040766-65040766 on Build GRCh38
rs147400066 Chr.14: 65040817-65040817 on Build GRCh38
rs768077835 Chr.14: 65040933-65040933 on Build GRCh38
rs898607911 Chr.14: 65040902-65040902 on Build GRCh38
rs184424696 Chr.14: 65040958-65040958 on Build GRCh38
rs145895625 Chr.14: 65040868-65040868 on Build GRCh38
rs200853756 Chr.14: 65040870-65040870 on Build GRCh38
rs1056465669 Chr.14: 65040822-65040822 on Build GRCh38
rs760464029 Chr.14: 65040871-65040871 on Build GRCh38
rs111476775 Chr.14: 65040940-65040940 on Build GRCh38
rs138149266 Chr.14: 65040820-65040820 on Build GRCh38
rs779620216 Chr.14: 65040970-65040973 on Build GRCh38
rs753303063 Chr.14: 65040765-65040765 on Build GRCh38
rs371596429 Chr.14: 65040808-65040808 on Build GRCh38
rs749947758 Chr.14: 65040894-65040894 on Build GRCh38
rs369967703 Chr.14: 65040968-65040968 on Build GRCh38
rs764889902 Chr.14: 65040886-65040886 on Build GRCh38
rs763790962 Chr.14: 65040744-65040744 on Build GRCh38
rs757830095 Chr.14: 65040952-65040952 on Build GRCh38
rs768199579 Chr.14: 65040874-65040874 on Build GRCh38
rs566646589 Chr.14: 65040901-65040901 on Build GRCh38
rs12588921 Chr.14: 65040789-65040789 on Build GRCh38
rs771996889 Chr.14: 65040869-65040869 on Build GRCh38

View Full Product Details