Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3785032-3785293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPB
Entrez Gene ID
1059
Gene Name
centromere protein B
Gene Aliases
-
UniGene
Hs.516855
Gene Chromosome Location
Chr.20: 3783851-3786690 on Build GRCh38
Transcripts
NM_001810.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280405 c.1334A>G p.E445G Chr.20: 3785150-3785150 on Build GRCh38
5745437 c.1448C>A p.A483D Chr.20: 3785036-3785036 on Build GRCh38
1200730 c.1380G>T p.E460D Chr.20: 3785104-3785104 on Build GRCh38
3291035 c.1339G>A p.E447K Chr.20: 3785145-3785145 on Build GRCh38
6525623 c.1259G>T p.G420V Chr.20: 3785225-3785225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761251384 Chr.20: 3785196-3785198 on Build GRCh38
rs745514430 Chr.20: 3785100-3785100 on Build GRCh38
rs754066175 Chr.20: 3785050-3785050 on Build GRCh38
rs996759685 Chr.20: 3785215-3785215 on Build GRCh38
rs780699266 Chr.20: 3785097-3785097 on Build GRCh38
rs745337300 Chr.20: 3785182-3785182 on Build GRCh38
rs748211874 Chr.20: 3785171-3785172 on Build GRCh38
rs773871246 Chr.20: 3785195-3785196 on Build GRCh38
rs752768027 Chr.20: 3785243-3785260 on Build GRCh38
rs199992333 Chr.20: 3785139-3785139 on Build GRCh38
rs1041906979 Chr.20: 3785069-3785069 on Build GRCh38
rs945775228 Chr.20: 3785054-3785054 on Build GRCh38
rs745612729 Chr.20: 3785180-3785182 on Build GRCh38
rs533227646 Chr.20: 3785145-3785145 on Build GRCh38
rs527821974 Chr.20: 3785176-3785176 on Build GRCh38
rs759706920 Chr.20: 3785134-3785134 on Build GRCh38
rs768120182 Chr.20: 3785185-3785186 on Build GRCh38
rs768121518 Chr.20: 3785247-3785247 on Build GRCh38
rs769640842 Chr.20: 3785179-3785180 on Build GRCh38
rs755554073 Chr.20: 3785185-3785185 on Build GRCh38
rs780860803 Chr.20: 3785155-3785155 on Build GRCh38
rs754455322 Chr.20: 3785099-3785101 on Build GRCh38
rs1036752785 Chr.20: 3785180-3785188 on Build GRCh38
rs529107942 Chr.20: 3785241-3785241 on Build GRCh38
rs755877996 Chr.20: 3785258-3785263 on Build GRCh38
rs757730302 Chr.20: 3785245-3785245 on Build GRCh38
rs756447571 Chr.20: 3785089-3785091 on Build GRCh38
rs563214391 Chr.20: 3785113-3785113 on Build GRCh38
rs144899160 Chr.20: 3785172-3785172 on Build GRCh38
rs772004115 Chr.20: 3785211-3785211 on Build GRCh38
rs761720824 Chr.20: 3785127-3785127 on Build GRCh38
rs755087750 Chr.20: 3785105-3785107 on Build GRCh38
rs774335332 Chr.20: 3785121-3785121 on Build GRCh38
rs201265512 Chr.20: 3785080-3785080 on Build GRCh38
rs142517860 Chr.20: 3785140-3785140 on Build GRCh38
rs367941845 Chr.20: 3785227-3785227 on Build GRCh38
rs947501798 Chr.20: 3785049-3785049 on Build GRCh38
rs778054791 Chr.20: 3785245-3785246 on Build GRCh38
rs748930909 Chr.20: 3785193-3785193 on Build GRCh38
rs779148447 Chr.20: 3785065-3785065 on Build GRCh38
rs768406399 Chr.20: 3785114-3785114 on Build GRCh38
rs755714252 Chr.20: 3785269-3785269 on Build GRCh38
rs6084465 Chr.20: 3785257-3785257 on Build GRCh38
rs757369944 Chr.20: 3785194-3785195 on Build GRCh38
rs560113368 Chr.20: 3785174-3785174 on Build GRCh38
rs777873622 Chr.20: 3785175-3785195 on Build GRCh38
rs774073734 Chr.20: 3785208-3785208 on Build GRCh38
rs769450213 Chr.20: 3785103-3785103 on Build GRCh38
rs779589417 Chr.20: 3785240-3785242 on Build GRCh38
rs755170043 Chr.20: 3785062-3785062 on Build GRCh38
rs750862703 Chr.20: 3785250-3785250 on Build GRCh38
rs752938535 Chr.20: 3785143-3785143 on Build GRCh38
rs779173611 Chr.20: 3785154-3785156 on Build GRCh38
rs746270700 Chr.20: 3785201-3785203 on Build GRCh38
rs748080776 Chr.20: 3785209-3785209 on Build GRCh38
rs766900282 Chr.20: 3785220-3785222 on Build GRCh38
rs200947984 Chr.20: 3785200-3785201 on Build GRCh38
rs780704260 Chr.20: 3785096-3785107 on Build GRCh38
rs780602403 Chr.20: 3785178-3785178 on Build GRCh38
rs768492671 Chr.20: 3785198-3785198 on Build GRCh38
rs747251087 Chr.20: 3785246-3785251 on Build GRCh38
rs138187623 Chr.20: 3785212-3785212 on Build GRCh38
rs545901224 Chr.20: 3785246-3785248 on Build GRCh38
rs761109209 Chr.20: 3785210-3785212 on Build GRCh38
rs755501841 Chr.20: 3785231-3785233 on Build GRCh38
rs779792835 Chr.20: 3785192-3785192 on Build GRCh38
rs779523200 Chr.20: 3785270-3785270 on Build GRCh38
rs759704025 Chr.20: 3785225-3785225 on Build GRCh38
rs772292203 Chr.20: 3785172-3785172 on Build GRCh38
rs754367212 Chr.20: 3785228-3785242 on Build GRCh38
rs750047148 Chr.20: 3785207-3785212 on Build GRCh38
rs145690729 Chr.20: 3785104-3785104 on Build GRCh38
rs765228310 Chr.20: 3785226-3785226 on Build GRCh38
rs767138027 Chr.20: 3785199-3785228 on Build GRCh38
rs747515195 Chr.20: 3785094-3785094 on Build GRCh38
rs756885952 Chr.20: 3785096-3785096 on Build GRCh38
rs762922762 Chr.20: 3785180-3785185 on Build GRCh38
rs191679255 Chr.20: 3785228-3785228 on Build GRCh38
rs373678745 Chr.20: 3785081-3785081 on Build GRCh38
rs983001182 Chr.20: 3785242-3785242 on Build GRCh38
rs13042058 Chr.20: 3785177-3785177 on Build GRCh38
rs138025965 Chr.20: 3785131-3785131 on Build GRCh38
rs551419159 Chr.20: 3785109-3785109 on Build GRCh38

View Full Product Details