Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111649525-111649766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111654351 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Gene Symbol
DNAJC25-GNG10
Entrez Gene ID
552891
Gene Name
DNAJC25-GNG10 readthrough
Gene Aliases
-
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111670246 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432407 c.742C>T p.Q248* Chr.9: 111649705-111649705 on Build GRCh38
1204210 c.607G>T p.E203* Chr.9: 111649570-111649570 on Build GRCh38
4163214 c.616A>T p.K206* Chr.9: 111649579-111649579 on Build GRCh38
3169976 c.799A>G p.I267V Chr.9: 111649762-111649762 on Build GRCh38
3169975 c.614G>A p.G205D Chr.9: 111649577-111649577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936093747 Chr.9: 111649565-111649565 on Build GRCh38
rs755601452 Chr.9: 111649566-111649566 on Build GRCh38
rs745714904 Chr.9: 111649735-111649735 on Build GRCh38
rs907133557 Chr.9: 111649706-111649706 on Build GRCh38
rs769980083 Chr.9: 111649618-111649618 on Build GRCh38
rs751963227 Chr.9: 111649576-111649576 on Build GRCh38
rs766749038 Chr.9: 111649647-111649647 on Build GRCh38
rs371744939 Chr.9: 111649636-111649636 on Build GRCh38
rs745598077 Chr.9: 111649606-111649606 on Build GRCh38
rs751987472 Chr.9: 111649655-111649655 on Build GRCh38
rs780670002 Chr.9: 111649551-111649551 on Build GRCh38
rs894828413 Chr.9: 111649663-111649663 on Build GRCh38
rs7019332 Chr.9: 111649665-111649665 on Build GRCh38
rs764455550 Chr.9: 111649570-111649570 on Build GRCh38
rs750062063 Chr.9: 111649684-111649684 on Build GRCh38
rs753283385 Chr.9: 111649728-111649728 on Build GRCh38
rs770703441 Chr.9: 111649572-111649572 on Build GRCh38
rs757237830 Chr.9: 111649729-111649729 on Build GRCh38
rs777750152 Chr.9: 111649569-111649569 on Build GRCh38
rs776960637 Chr.9: 111649610-111649610 on Build GRCh38
rs376569149 Chr.9: 111649580-111649580 on Build GRCh38
rs778671787 Chr.9: 111649731-111649731 on Build GRCh38
rs762032954 Chr.9: 111649612-111649612 on Build GRCh38
rs868430633 Chr.9: 111649672-111649672 on Build GRCh38
rs768953133 Chr.9: 111649607-111649607 on Build GRCh38
rs755691509 Chr.9: 111649688-111649688 on Build GRCh38
rs752247206 Chr.9: 111649559-111649559 on Build GRCh38
rs766270759 Chr.9: 111649612-111649614 on Build GRCh38
rs777266187 Chr.9: 111649698-111649698 on Build GRCh38
rs930948272 Chr.9: 111649621-111649621 on Build GRCh38
rs763529345 Chr.9: 111649642-111649642 on Build GRCh38
rs1322259 Chr.9: 111649611-111649611 on Build GRCh38
rs534213065 Chr.9: 111649658-111649658 on Build GRCh38
rs773913181 Chr.9: 111649589-111649589 on Build GRCh38

View Full Product Details