Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86604811-86605048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374845
Entrez Gene ID
105374845
Gene Name
uncharacterized LOC105374845
Gene Aliases
-
UniGene
Hs.741633
Gene Chromosome Location
Chr.2: 86562345-86618766 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103
Entrez Gene ID
7844
Gene Name
ring finger protein 103
Gene Aliases
HKF-1, KF-1, KF1, ZFP-103, ZFP103
UniGene
Hs.469199
Gene Chromosome Location
Chr.2: 86603393-86623877 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103-CHMP3
Entrez Gene ID
100526767
Gene Name
RNF103-CHMP3 readthrough
Gene Aliases
RNF103-VPS24
UniGene
Hs.591582
Gene Chromosome Location
Chr.2: 86503430-86721122 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3799255 c.953C>G p.S318* Chr.2: 86604948-86604948 on Build GRCh38
4583684 c.1084A>G p.T362A Chr.2: 86604817-86604817 on Build GRCh38
6402387 c.908C>T p.T303I Chr.2: 86604993-86604993 on Build GRCh38
5025668 c.1040G>A p.G347E Chr.2: 86604861-86604861 on Build GRCh38
3735194 c.941C>A p.S314* Chr.2: 86604960-86604960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754426665 Chr.2: 86604992-86604992 on Build GRCh38
rs756645428 Chr.2: 86604842-86604842 on Build GRCh38
rs755509760 Chr.2: 86604995-86604995 on Build GRCh38
rs770659862 Chr.2: 86604938-86604938 on Build GRCh38
rs201780922 Chr.2: 86604958-86604958 on Build GRCh38
rs370678832 Chr.2: 86605014-86605014 on Build GRCh38
rs531471942 Chr.2: 86604941-86604941 on Build GRCh38
rs749749696 Chr.2: 86604851-86604851 on Build GRCh38
rs759429482 Chr.2: 86604950-86604950 on Build GRCh38
rs774394413 Chr.2: 86604961-86604961 on Build GRCh38
rs762137122 Chr.2: 86604930-86604930 on Build GRCh38
rs750543175 Chr.2: 86604986-86604986 on Build GRCh38
rs772950406 Chr.2: 86604885-86604885 on Build GRCh38
rs746828990 Chr.2: 86604908-86604908 on Build GRCh38
rs766796913 Chr.2: 86604989-86604989 on Build GRCh38
rs147773943 Chr.2: 86605004-86605004 on Build GRCh38
rs750784881 Chr.2: 86604841-86604841 on Build GRCh38
rs989413693 Chr.2: 86604952-86604952 on Build GRCh38
rs202140822 Chr.2: 86604863-86604863 on Build GRCh38
rs780710494 Chr.2: 86604846-86604846 on Build GRCh38
rs996207000 Chr.2: 86604871-86604871 on Build GRCh38
rs958293718 Chr.2: 86604892-86604892 on Build GRCh38
rs754553526 Chr.2: 86604853-86604853 on Build GRCh38
rs778613314 Chr.2: 86604996-86604996 on Build GRCh38
rs61760875 Chr.2: 86605009-86605009 on Build GRCh38
rs765625023 Chr.2: 86604937-86604937 on Build GRCh38
rs757781314 Chr.2: 86605010-86605010 on Build GRCh38
rs767773623 Chr.2: 86604981-86604981 on Build GRCh38
rs747792563 Chr.2: 86604866-86604866 on Build GRCh38
rs771744537 Chr.2: 86604874-86604874 on Build GRCh38
rs147608406 Chr.2: 86604980-86604980 on Build GRCh38

View Full Product Details