Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15436145-15436388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF1
Entrez Gene ID
85403
Gene Name
ELL associated factor 1
Gene Aliases
-
UniGene
Hs.716733
Gene Chromosome Location
Chr.3: 15427557-15442613 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1 XM_011534165.1 XM_011534166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327791 c.548T>G p.I183S Chr.3: 15436363-15436363 on Build GRCh38
1192952 c.561_562insC p.S188fs*12 Chr.3: 15436376-15436377 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749754070 Chr.3: 15436319-15436319 on Build GRCh38
rs532966683 Chr.3: 15436181-15436181 on Build GRCh38
rs535493318 Chr.3: 15436366-15436366 on Build GRCh38
rs771523869 Chr.3: 15436336-15436336 on Build GRCh38
rs748458745 Chr.3: 15436310-15436310 on Build GRCh38
rs547770251 Chr.3: 15436241-15436241 on Build GRCh38
rs890748644 Chr.3: 15436220-15436220 on Build GRCh38
rs774271370 Chr.3: 15436346-15436346 on Build GRCh38
rs780748188 Chr.3: 15436196-15436196 on Build GRCh38
rs1042009432 Chr.3: 15436168-15436173 on Build GRCh38
rs1012823042 Chr.3: 15436197-15436197 on Build GRCh38
rs778184743 Chr.3: 15436314-15436314 on Build GRCh38
rs971285866 Chr.3: 15436254-15436254 on Build GRCh38
rs754591118 Chr.3: 15436306-15436306 on Build GRCh38
rs772386193 Chr.3: 15436367-15436367 on Build GRCh38
rs780851127 Chr.3: 15436307-15436307 on Build GRCh38
rs957275283 Chr.3: 15436169-15436169 on Build GRCh38
rs549965906 Chr.3: 15436186-15436186 on Build GRCh38
rs370763713 Chr.3: 15436323-15436323 on Build GRCh38
rs770153308 Chr.3: 15436311-15436311 on Build GRCh38
rs374684025 Chr.3: 15436215-15436215 on Build GRCh38
rs138937059 Chr.3: 15436286-15436286 on Build GRCh38
rs950692079 Chr.3: 15436355-15436355 on Build GRCh38

View Full Product Details