Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33679682-33679955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857692 c.454G>A p.D152N Chr.20: 33679873-33679873 on Build GRCh38
244178 c.380G>A p.R127H Chr.20: 33679947-33679947 on Build GRCh38
5900640 c.496C>T p.R166C Chr.20: 33679831-33679831 on Build GRCh38
5955040 c.571C>T p.L191L Chr.20: 33679756-33679756 on Build GRCh38
6004566 c.451G>A p.V151I Chr.20: 33679876-33679876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377425346 Chr.20: 33679789-33679789 on Build GRCh38
rs374343061 Chr.20: 33679730-33679730 on Build GRCh38
rs756301576 Chr.20: 33679738-33679739 on Build GRCh38
rs201315516 Chr.20: 33679835-33679835 on Build GRCh38
rs373698861 Chr.20: 33679874-33679874 on Build GRCh38
rs777413332 Chr.20: 33679721-33679721 on Build GRCh38
rs371120550 Chr.20: 33679852-33679852 on Build GRCh38
rs560667741 Chr.20: 33679715-33679715 on Build GRCh38
rs752291306 Chr.20: 33679875-33679875 on Build GRCh38
rs769189668 Chr.20: 33679746-33679746 on Build GRCh38
rs754145642 Chr.20: 33679923-33679924 on Build GRCh38
rs756189133 Chr.20: 33679745-33679745 on Build GRCh38
rs780712505 Chr.20: 33679738-33679738 on Build GRCh38
rs997770717 Chr.20: 33679877-33679877 on Build GRCh38
rs760671739 Chr.20: 33679829-33679829 on Build GRCh38
rs751015434 Chr.20: 33679887-33679887 on Build GRCh38
rs746347817 Chr.20: 33679729-33679729 on Build GRCh38
rs755388533 Chr.20: 33679923-33679923 on Build GRCh38
rs759306871 Chr.20: 33679855-33679855 on Build GRCh38
rs766261474 Chr.20: 33679847-33679847 on Build GRCh38
rs749851572 Chr.20: 33679919-33679919 on Build GRCh38
rs780682077 Chr.20: 33679900-33679900 on Build GRCh38
rs201080895 Chr.20: 33679901-33679901 on Build GRCh38
rs907212730 Chr.20: 33679724-33679724 on Build GRCh38
rs752446932 Chr.20: 33679708-33679708 on Build GRCh38
rs1020536486 Chr.20: 33679817-33679817 on Build GRCh38
rs146062687 Chr.20: 33679805-33679805 on Build GRCh38
rs864622017 Chr.20: 33679830-33679830 on Build GRCh38
rs763667605 Chr.20: 33679883-33679883 on Build GRCh38
rs139667011 Chr.20: 33679811-33679811 on Build GRCh38
rs750229586 Chr.20: 33679742-33679742 on Build GRCh38
rs751400403 Chr.20: 33679723-33679723 on Build GRCh38
rs200021458 Chr.20: 33679889-33679889 on Build GRCh38
rs753403393 Chr.20: 33679922-33679922 on Build GRCh38
rs1020522202 Chr.20: 33679727-33679727 on Build GRCh38
rs758127423 Chr.20: 33679876-33679876 on Build GRCh38
rs780399809 Chr.20: 33679809-33679810 on Build GRCh38
rs773281069 Chr.20: 33679820-33679820 on Build GRCh38
rs202180598 Chr.20: 33679737-33679737 on Build GRCh38
rs903398417 Chr.20: 33679856-33679856 on Build GRCh38

View Full Product Details