Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83030183-83030422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
UniGene
Hs.459149
Gene Chromosome Location
Chr.15: 83016423-83067434 on Build GRCh38
Transcripts
NM_001011885.1 NM_025238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39882 c.978G>A p.R326R Chr.15: 83030213-83030213 on Build GRCh38
6226759 c.875C>T p.S292F Chr.15: 83030316-83030316 on Build GRCh38
965810 c.1006C>T p.Q336* Chr.15: 83030185-83030185 on Build GRCh38
6613293 c.958C>T p.R320* Chr.15: 83030233-83030233 on Build GRCh38
965811 c.941G>A p.R314Q Chr.15: 83030250-83030250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367774760 Chr.15: 83030247-83030247 on Build GRCh38
rs762382059 Chr.15: 83030276-83030276 on Build GRCh38
rs761617843 Chr.15: 83030238-83030238 on Build GRCh38
rs756563348 Chr.15: 83030253-83030253 on Build GRCh38
rs751620814 Chr.15: 83030362-83030362 on Build GRCh38
rs115684450 Chr.15: 83030290-83030290 on Build GRCh38
rs138518925 Chr.15: 83030233-83030233 on Build GRCh38
rs1024878284 Chr.15: 83030292-83030292 on Build GRCh38
rs771795964 Chr.15: 83030351-83030351 on Build GRCh38
rs574934563 Chr.15: 83030229-83030229 on Build GRCh38
rs770405819 Chr.15: 83030279-83030280 on Build GRCh38
rs745612106 Chr.15: 83030309-83030309 on Build GRCh38
rs144118813 Chr.15: 83030308-83030308 on Build GRCh38
rs773905310 Chr.15: 83030352-83030353 on Build GRCh38
rs559906250 Chr.15: 83030372-83030372 on Build GRCh38
rs748394929 Chr.15: 83030295-83030295 on Build GRCh38
rs766863988 Chr.15: 83030258-83030258 on Build GRCh38
rs1009237736 Chr.15: 83030357-83030357 on Build GRCh38
rs769607961 Chr.15: 83030316-83030316 on Build GRCh38
rs577288866 Chr.15: 83030298-83030298 on Build GRCh38
rs116584930 Chr.15: 83030216-83030216 on Build GRCh38
rs754338254 Chr.15: 83030281-83030281 on Build GRCh38
rs751514037 Chr.15: 83030299-83030299 on Build GRCh38
rs375845805 Chr.15: 83030232-83030232 on Build GRCh38
rs768200976 Chr.15: 83030331-83030331 on Build GRCh38
rs780011156 Chr.15: 83030326-83030326 on Build GRCh38
rs372203136 Chr.15: 83030291-83030291 on Build GRCh38
rs749233777 Chr.15: 83030329-83030329 on Build GRCh38
rs761084676 Chr.15: 83030365-83030365 on Build GRCh38
rs750752981 Chr.15: 83030251-83030251 on Build GRCh38
rs374731662 Chr.15: 83030350-83030350 on Build GRCh38
rs754353864 Chr.15: 83030377-83030377 on Build GRCh38

View Full Product Details