Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28717808-28718077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6213304 c.1848C>T p.H616H Chr.8: 28717907-28717907 on Build GRCh38
7255720 c.1758A>T p.S586S Chr.8: 28717817-28717817 on Build GRCh38
5351506 c.1853C>T p.P618L Chr.8: 28717912-28717912 on Build GRCh38
6180715 c.1771C>A p.R591S Chr.8: 28717830-28717830 on Build GRCh38
109076 c.1770C>T p.L590L Chr.8: 28717829-28717829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752466548 Chr.8: 28717964-28717964 on Build GRCh38
rs749822351 Chr.8: 28717893-28717893 on Build GRCh38
rs376886406 Chr.8: 28718051-28718051 on Build GRCh38
rs376869552 Chr.8: 28717831-28717831 on Build GRCh38
rs746518185 Chr.8: 28717978-28717978 on Build GRCh38
rs754585925 Chr.8: 28717958-28717958 on Build GRCh38
rs775682299 Chr.8: 28718018-28718018 on Build GRCh38
rs751064521 Chr.8: 28717957-28717957 on Build GRCh38
rs201439713 Chr.8: 28717828-28717828 on Build GRCh38
rs1050016206 Chr.8: 28717881-28717881 on Build GRCh38
rs771306625 Chr.8: 28718001-28718001 on Build GRCh38
rs765007284 Chr.8: 28717931-28717931 on Build GRCh38
rs775735115 Chr.8: 28717919-28717919 on Build GRCh38
rs766397327 Chr.8: 28718052-28718052 on Build GRCh38
rs770098386 Chr.8: 28717863-28717863 on Build GRCh38
rs745922565 Chr.8: 28718014-28718014 on Build GRCh38
rs777945396 Chr.8: 28717974-28717974 on Build GRCh38
rs368676648 Chr.8: 28718002-28718002 on Build GRCh38
rs891358223 Chr.8: 28717886-28717886 on Build GRCh38
rs200563165 Chr.8: 28717896-28717896 on Build GRCh38
rs147333542 Chr.8: 28717829-28717829 on Build GRCh38
rs979882136 Chr.8: 28718046-28718046 on Build GRCh38
rs766423907 Chr.8: 28717953-28717953 on Build GRCh38
rs891789506 Chr.8: 28717920-28717920 on Build GRCh38
rs749670990 Chr.8: 28717991-28717991 on Build GRCh38
rs373139263 Chr.8: 28718015-28718015 on Build GRCh38
rs888609307 Chr.8: 28718038-28718038 on Build GRCh38
rs754610555 Chr.8: 28717840-28717840 on Build GRCh38
rs768934948 Chr.8: 28718035-28718035 on Build GRCh38
rs747753901 Chr.8: 28717909-28717909 on Build GRCh38
rs770842408 Chr.8: 28718029-28718029 on Build GRCh38
rs143652582 Chr.8: 28717934-28717934 on Build GRCh38
rs937010739 Chr.8: 28717923-28717923 on Build GRCh38
rs969877872 Chr.8: 28718037-28718037 on Build GRCh38
rs771484952 Chr.8: 28717894-28717894 on Build GRCh38
rs756490197 Chr.8: 28717969-28717969 on Build GRCh38
rs746530379 Chr.8: 28717835-28717835 on Build GRCh38
rs762837299 Chr.8: 28718050-28718050 on Build GRCh38
rs780962047 Chr.8: 28717963-28717963 on Build GRCh38
rs1042826033 Chr.8: 28717842-28717842 on Build GRCh38
rs762741027 Chr.8: 28717947-28717947 on Build GRCh38
rs781014999 Chr.8: 28717852-28717852 on Build GRCh38
rs759647888 Chr.8: 28717901-28717901 on Build GRCh38
rs761635451 Chr.8: 28717925-28717925 on Build GRCh38
rs773766888 Chr.8: 28717864-28717864 on Build GRCh38
rs138175937 Chr.8: 28718036-28718036 on Build GRCh38
rs748502441 Chr.8: 28717862-28717862 on Build GRCh38
rs777350737 Chr.8: 28717968-28717968 on Build GRCh38
rs771738641 Chr.8: 28717914-28717914 on Build GRCh38
rs996983305 Chr.8: 28717850-28717850 on Build GRCh38

View Full Product Details