Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75042094-75042365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5H
Entrez Gene ID
10476
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Gene Aliases
ATPQ
UniGene
Hs.514465
Gene Chromosome Location
Chr.17: 75038860-75046979 on Build GRCh38
Transcripts
NM_001003785.1 NM_006356.2 NR_110835.1

Gene Symbol
KCTD2
Entrez Gene ID
23510
Gene Name
potassium channel tetramerization domain containing 2
Gene Aliases
-
UniGene
Hs.514468
Gene Chromosome Location
Chr.17: 75032567-75065889 on Build GRCh38
Transcripts
NM_001003785.1 NM_006356.2 NR_110835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937557 c.127G>T p.A43S Chr.17: 75042273-75042273 on Build GRCh38
6607958 c.156C>T p.I52I Chr.17: 75042244-75042244 on Build GRCh38
6430337 c.184G>A p.A62T Chr.17: 75042216-75042216 on Build GRCh38
6428851 c.212A>T p.E71V Chr.17: 75042188-75042188 on Build GRCh38
5813047 c.164C>A p.A55D Chr.17: 75042236-75042236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377594522 Chr.17: 75042243-75042243 on Build GRCh38
rs778317230 Chr.17: 75042266-75042266 on Build GRCh38
rs375365983 Chr.17: 75042289-75042289 on Build GRCh38
rs777346570 Chr.17: 75042205-75042205 on Build GRCh38
rs141535456 Chr.17: 75042253-75042253 on Build GRCh38
rs377590780 Chr.17: 75042244-75042244 on Build GRCh38
rs1035771941 Chr.17: 75042290-75042290 on Build GRCh38
rs965130437 Chr.17: 75042319-75042319 on Build GRCh38
rs201437197 Chr.17: 75042184-75042184 on Build GRCh38
rs553483525 Chr.17: 75042246-75042246 on Build GRCh38
rs147284668 Chr.17: 75042224-75042224 on Build GRCh38
rs755493665 Chr.17: 75042152-75042152 on Build GRCh38
rs371524705 Chr.17: 75042327-75042327 on Build GRCh38
rs138253735 Chr.17: 75042215-75042215 on Build GRCh38
rs200793762 Chr.17: 75042249-75042249 on Build GRCh38
rs768267752 Chr.17: 75042296-75042297 on Build GRCh38
rs375367412 Chr.17: 75042285-75042285 on Build GRCh38
rs751878822 Chr.17: 75042150-75042150 on Build GRCh38
rs752152162 Chr.17: 75042162-75042162 on Build GRCh38
rs767950776 Chr.17: 75042157-75042157 on Build GRCh38
rs746722852 Chr.17: 75042181-75042183 on Build GRCh38
rs778434521 Chr.17: 75042142-75042142 on Build GRCh38
rs774708814 Chr.17: 75042282-75042282 on Build GRCh38
rs143915019 Chr.17: 75042221-75042221 on Build GRCh38
rs763287590 Chr.17: 75042146-75042146 on Build GRCh38
rs1017912378 Chr.17: 75042309-75042309 on Build GRCh38
rs1001616314 Chr.17: 75042287-75042287 on Build GRCh38
rs190471145 Chr.17: 75042278-75042278 on Build GRCh38
rs776152275 Chr.17: 75042328-75042341 on Build GRCh38
rs765385788 Chr.17: 75042137-75042137 on Build GRCh38
rs745611418 Chr.17: 75042226-75042226 on Build GRCh38
rs376934365 Chr.17: 75042286-75042286 on Build GRCh38
rs777678768 Chr.17: 75042329-75042329 on Build GRCh38
rs772149859 Chr.17: 75042241-75042241 on Build GRCh38
rs200283373 Chr.17: 75042276-75042276 on Build GRCh38
rs763264238 Chr.17: 75042259-75042259 on Build GRCh38
rs960368245 Chr.17: 75042308-75042308 on Build GRCh38
rs369589395 Chr.17: 75042148-75042148 on Build GRCh38

View Full Product Details