Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15577650-15577901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMAT
Entrez Gene ID
79814
Gene Name
agmatinase
Gene Aliases
-
UniGene
Hs.461532
Gene Chromosome Location
Chr.1: 15571699-15585110 on Build GRCh38
Transcripts
NM_024758.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334198 c.871C>A p.P291T Chr.1: 15577714-15577714 on Build GRCh38
3965935 c.849C>T p.A283A Chr.1: 15577736-15577736 on Build GRCh38
5914536 c.769C>T p.P257S Chr.1: 15577816-15577816 on Build GRCh38
6875572 c.742G>A p.E248K Chr.1: 15577843-15577843 on Build GRCh38
3476413 c.840G>T p.L280L Chr.1: 15577745-15577745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755628910 Chr.1: 15577857-15577857 on Build GRCh38
rs148750290 Chr.1: 15577751-15577751 on Build GRCh38
rs773741199 Chr.1: 15577765-15577765 on Build GRCh38
rs756846132 Chr.1: 15577834-15577834 on Build GRCh38
rs766717409 Chr.1: 15577788-15577788 on Build GRCh38
rs971074936 Chr.1: 15577865-15577865 on Build GRCh38
rs772726449 Chr.1: 15577875-15577875 on Build GRCh38
rs895965250 Chr.1: 15577824-15577824 on Build GRCh38
rs758187940 Chr.1: 15577714-15577714 on Build GRCh38
rs754883011 Chr.1: 15577695-15577695 on Build GRCh38
rs947372532 Chr.1: 15577678-15577678 on Build GRCh38
rs777384893 Chr.1: 15577727-15577727 on Build GRCh38
rs11544523 Chr.1: 15577823-15577823 on Build GRCh38
rs757702716 Chr.1: 15577843-15577843 on Build GRCh38
rs753169965 Chr.1: 15577801-15577802 on Build GRCh38
rs752628452 Chr.1: 15577811-15577811 on Build GRCh38
rs760928663 Chr.1: 15577779-15577779 on Build GRCh38
rs748559540 Chr.1: 15577864-15577864 on Build GRCh38
rs138227623 Chr.1: 15577858-15577858 on Build GRCh38
rs773649439 Chr.1: 15577877-15577877 on Build GRCh38
rs766524915 Chr.1: 15577680-15577680 on Build GRCh38
rs765361154 Chr.1: 15577671-15577672 on Build GRCh38
rs759604775 Chr.1: 15577806-15577806 on Build GRCh38
rs770602965 Chr.1: 15577729-15577729 on Build GRCh38
rs769200724 Chr.1: 15577732-15577732 on Build GRCh38
rs977516649 Chr.1: 15577872-15577872 on Build GRCh38
rs780632221 Chr.1: 15577730-15577730 on Build GRCh38
rs745384069 Chr.1: 15577731-15577731 on Build GRCh38
rs760803794 Chr.1: 15577673-15577673 on Build GRCh38
rs772603937 Chr.1: 15577752-15577752 on Build GRCh38
rs745370110 Chr.1: 15577856-15577856 on Build GRCh38
rs753881523 Chr.1: 15577693-15577693 on Build GRCh38
rs758160748 Chr.1: 15577816-15577816 on Build GRCh38
rs991438754 Chr.1: 15577852-15577852 on Build GRCh38
rs752536854 Chr.1: 15577702-15577702 on Build GRCh38
rs778724560 Chr.1: 15577696-15577696 on Build GRCh38
rs765212237 Chr.1: 15577807-15577807 on Build GRCh38
rs962159105 Chr.1: 15577747-15577747 on Build GRCh38
rs370519006 Chr.1: 15577802-15577802 on Build GRCh38
rs149178480 Chr.1: 15577826-15577826 on Build GRCh38
rs774777432 Chr.1: 15577750-15577750 on Build GRCh38
rs148211163 Chr.1: 15577836-15577836 on Build GRCh38
rs974374334 Chr.1: 15577777-15577777 on Build GRCh38

View Full Product Details