Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106239914-106240185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP4
Entrez Gene ID
10970
Gene Name
cytoskeleton associated protein 4
Gene Aliases
CLIMP-63, ERGIC-63, p63
UniGene
Hs.74368
Gene Chromosome Location
Chr.12: 106237881-106247935 on Build GRCh38
Transcripts
NM_006825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629533 c.720G>A p.R240R Chr.12: 106240113-106240113 on Build GRCh38
5653706 c.867A>T p.L289F Chr.12: 106239966-106239966 on Build GRCh38
4038259 c.692C>T p.T231M Chr.12: 106240141-106240141 on Build GRCh38
6629531 c.786C>T p.S262S Chr.12: 106240047-106240047 on Build GRCh38
1944928 c.672C>T p.D224D Chr.12: 106240161-106240161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143701933 Chr.12: 106240032-106240032 on Build GRCh38
rs767034363 Chr.12: 106240160-106240160 on Build GRCh38
rs755671922 Chr.12: 106240089-106240089 on Build GRCh38
rs766734379 Chr.12: 106240076-106240076 on Build GRCh38
rs773634121 Chr.12: 106240124-106240124 on Build GRCh38
rs781761637 Chr.12: 106240041-106240043 on Build GRCh38
rs369412968 Chr.12: 106240001-106240001 on Build GRCh38
rs115785750 Chr.12: 106240120-106240120 on Build GRCh38
rs778981244 Chr.12: 106240107-106240107 on Build GRCh38
rs752322663 Chr.12: 106240087-106240087 on Build GRCh38
rs754476378 Chr.12: 106240085-106240085 on Build GRCh38
rs374201449 Chr.12: 106240074-106240074 on Build GRCh38
rs747998993 Chr.12: 106239966-106239966 on Build GRCh38
rs776989409 Chr.12: 106240050-106240050 on Build GRCh38
rs199654199 Chr.12: 106240109-106240109 on Build GRCh38
rs201448750 Chr.12: 106240161-106240161 on Build GRCh38
rs775643945 Chr.12: 106240028-106240028 on Build GRCh38
rs988969873 Chr.12: 106239970-106239970 on Build GRCh38
rs765615556 Chr.12: 106240060-106240060 on Build GRCh38
rs777044508 Chr.12: 106240112-106240112 on Build GRCh38
rs748427432 Chr.12: 106240115-106240115 on Build GRCh38
rs369547770 Chr.12: 106240077-106240077 on Build GRCh38
rs774804936 Chr.12: 106240150-106240150 on Build GRCh38
rs766972254 Chr.12: 106240136-106240136 on Build GRCh38
rs762309036 Chr.12: 106240052-106240052 on Build GRCh38
rs202223849 Chr.12: 106240079-106240079 on Build GRCh38
rs144144747 Chr.12: 106240108-106240108 on Build GRCh38
rs770732132 Chr.12: 106240017-106240017 on Build GRCh38
rs921094257 Chr.12: 106240016-106240016 on Build GRCh38
rs757231622 Chr.12: 106240101-106240101 on Build GRCh38
rs150849660 Chr.12: 106240156-106240156 on Build GRCh38
rs12818106 Chr.12: 106239939-106239939 on Build GRCh38
rs141436009 Chr.12: 106240125-106240125 on Build GRCh38
rs764382147 Chr.12: 106240151-106240151 on Build GRCh38
rs899320767 Chr.12: 106240141-106240141 on Build GRCh38
rs748968259 Chr.12: 106240097-106240097 on Build GRCh38
rs1043334333 Chr.12: 106240025-106240025 on Build GRCh38
rs769058661 Chr.12: 106240035-106240035 on Build GRCh38
rs199595372 Chr.12: 106240086-106240086 on Build GRCh38
rs377249485 Chr.12: 106239953-106239953 on Build GRCh38
rs773402608 Chr.12: 106240065-106240065 on Build GRCh38
rs377084647 Chr.12: 106240096-106240096 on Build GRCh38
rs755904848 Chr.12: 106239974-106239974 on Build GRCh38
rs777617893 Chr.12: 106239980-106239980 on Build GRCh38

View Full Product Details