Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125995358-125995599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANK1
Entrez Gene ID
92565
Gene Name
fibronectin type III and ankyrin repeat domains 1
Gene Aliases
HSD13
UniGene
Hs.352591
Gene Chromosome Location
Chr.10: 125896539-126009599 on Build GRCh38
Transcripts
NM_145235.3 XM_005270280.3 XM_006718058.3 XM_006718064.3 XM_011540348.2 XM_011540349.2 XM_011540350.2 XM_011540351.2 XM_011540356.2 XM_011540357.2 XM_011540358.2 XM_011540360.2 XM_011540361.2 XM_011540362.2 XM_011540363.2 XM_017016916.1 XM_017016917.1 XM_017016918.1 XM_017016919.1 XM_017016920.1 XM_017016921.1 XM_017016922.1 XM_017016923.1 XM_017016924.1 XM_017016925.1 XM_017016926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5660413 c.357T>C p.S119S Chr.10: 125995457-125995457 on Build GRCh38
4799231 c.322C>T p.P108S Chr.10: 125995422-125995422 on Build GRCh38
5919871 c.382C>G p.R128G Chr.10: 125995482-125995482 on Build GRCh38
6483743 c.398G>A p.G133D Chr.10: 125995498-125995498 on Build GRCh38
4761156 c.325A>T p.I109L Chr.10: 125995425-125995425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760423814 Chr.10: 125995533-125995533 on Build GRCh38
rs144707515 Chr.10: 125995460-125995460 on Build GRCh38
rs763552659 Chr.10: 125995407-125995407 on Build GRCh38
rs551850468 Chr.10: 125995494-125995494 on Build GRCh38
rs746141002 Chr.10: 125995482-125995482 on Build GRCh38
rs376761181 Chr.10: 125995385-125995385 on Build GRCh38
rs868261343 Chr.10: 125995551-125995551 on Build GRCh38
rs977297678 Chr.10: 125995559-125995559 on Build GRCh38
rs775866249 Chr.10: 125995485-125995485 on Build GRCh38
rs761467736 Chr.10: 125995420-125995420 on Build GRCh38
rs769052228 Chr.10: 125995519-125995519 on Build GRCh38
rs372993644 Chr.10: 125995467-125995467 on Build GRCh38
rs763476737 Chr.10: 125995405-125995405 on Build GRCh38
rs116737503 Chr.10: 125995555-125995555 on Build GRCh38
rs770187871 Chr.10: 125995381-125995381 on Build GRCh38
rs1019634886 Chr.10: 125995570-125995570 on Build GRCh38
rs954997862 Chr.10: 125995466-125995466 on Build GRCh38
rs761224815 Chr.10: 125995526-125995526 on Build GRCh38
rs202054807 Chr.10: 125995447-125995447 on Build GRCh38
rs147888723 Chr.10: 125995483-125995483 on Build GRCh38
rs753777352 Chr.10: 125995445-125995445 on Build GRCh38
rs767093060 Chr.10: 125995528-125995528 on Build GRCh38
rs768176029 Chr.10: 125995520-125995520 on Build GRCh38
rs370543774 Chr.10: 125995558-125995558 on Build GRCh38
rs750156778 Chr.10: 125995425-125995425 on Build GRCh38
rs754878501 Chr.10: 125995546-125995546 on Build GRCh38
rs751996540 Chr.10: 125995478-125995478 on Build GRCh38
rs894508240 Chr.10: 125995406-125995406 on Build GRCh38
rs755940888 Chr.10: 125995431-125995431 on Build GRCh38
rs924408886 Chr.10: 125995563-125995563 on Build GRCh38
rs750662286 Chr.10: 125995421-125995421 on Build GRCh38
rs753701348 Chr.10: 125995545-125995545 on Build GRCh38
rs975069208 Chr.10: 125995553-125995553 on Build GRCh38
rs141007393 Chr.10: 125995446-125995446 on Build GRCh38
rs1037789199 Chr.10: 125995432-125995432 on Build GRCh38
rs199625748 Chr.10: 125995417-125995417 on Build GRCh38
rs200785168 Chr.10: 125995519-125995520 on Build GRCh38
rs750958774 Chr.10: 125995541-125995542 on Build GRCh38
rs780308807 Chr.10: 125995539-125995539 on Build GRCh38
rs766323993 Chr.10: 125995441-125995441 on Build GRCh38
rs774976820 Chr.10: 125995525-125995525 on Build GRCh38
rs201725687 Chr.10: 125995532-125995532 on Build GRCh38
rs747047013 Chr.10: 125995455-125995455 on Build GRCh38
rs954827421 Chr.10: 125995541-125995541 on Build GRCh38

View Full Product Details