Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84971345-84971574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCMF1
Entrez Gene ID
56888
Gene Name
potassium channel modulatory factor 1
Gene Aliases
DEBT91, FIGC, PCMF, ZZZ1
UniGene
Hs.654968
Gene Chromosome Location
Chr.2: 84971108-85059472 on Build GRCh38
Transcripts
NM_020122.4

SNP Detail

SNP ID Chromosome Location
rs1014509710 Chr.2: 84971552-84971552 on Build GRCh38
rs762014929 Chr.2: 84971499-84971519 on Build GRCh38
rs1049120718 Chr.2: 84971518-84971518 on Build GRCh38
rs979769474 Chr.2: 84971367-84971367 on Build GRCh38
rs935775141 Chr.2: 84971370-84971370 on Build GRCh38
rs887845192 Chr.2: 84971522-84971522 on Build GRCh38
rs777017023 Chr.2: 84971492-84971493 on Build GRCh38
rs1053296268 Chr.2: 84971379-84971379 on Build GRCh38
rs996172251 Chr.2: 84971504-84971504 on Build GRCh38
rs539655134 Chr.2: 84971415-84971415 on Build GRCh38
rs762114426 Chr.2: 84971478-84971478 on Build GRCh38
rs1002145722 Chr.2: 84971532-84971552 on Build GRCh38
rs552044062 Chr.2: 84971493-84971493 on Build GRCh38
rs756384386 Chr.2: 84971517-84971517 on Build GRCh38
rs1015438512 Chr.2: 84971532-84971532 on Build GRCh38
rs913370555 Chr.2: 84971393-84971393 on Build GRCh38
rs759552848 Chr.2: 84971419-84971419 on Build GRCh38
rs764239820 Chr.2: 84971424-84971424 on Build GRCh38
rs774624300 Chr.2: 84971441-84971441 on Build GRCh38
rs776386097 Chr.2: 84971407-84971407 on Build GRCh38
rs926115842 Chr.2: 84971401-84971401 on Build GRCh38
rs961195146 Chr.2: 84971535-84971535 on Build GRCh38
rs969691001 Chr.2: 84971366-84971366 on Build GRCh38
rs767918601 Chr.2: 84971488-84971488 on Build GRCh38

View Full Product Details