Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86604611-86604882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374845
Entrez Gene ID
105374845
Gene Name
uncharacterized LOC105374845
Gene Aliases
-
UniGene
Hs.741633
Gene Chromosome Location
Chr.2: 86562345-86618766 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103
Entrez Gene ID
7844
Gene Name
ring finger protein 103
Gene Aliases
HKF-1, KF-1, KF1, ZFP-103, ZFP103
UniGene
Hs.469199
Gene Chromosome Location
Chr.2: 86603393-86623877 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103-CHMP3
Entrez Gene ID
100526767
Gene Name
RNF103-CHMP3 readthrough
Gene Aliases
RNF103-VPS24
UniGene
Hs.591582
Gene Chromosome Location
Chr.2: 86503430-86721122 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380284 c.1216G>A p.V406I Chr.2: 86604685-86604685 on Build GRCh38
5770905 c.1096C>T p.L366L Chr.2: 86604805-86604805 on Build GRCh38
1690764 c.1109C>T p.S370F Chr.2: 86604792-86604792 on Build GRCh38
3426743 c.1101A>C p.L367F Chr.2: 86604800-86604800 on Build GRCh38
4583684 c.1084A>G p.T362A Chr.2: 86604817-86604817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756645428 Chr.2: 86604842-86604842 on Build GRCh38
rs936164975 Chr.2: 86604773-86604773 on Build GRCh38
rs771790880 Chr.2: 86604764-86604764 on Build GRCh38
rs369237335 Chr.2: 86604796-86604796 on Build GRCh38
rs779542435 Chr.2: 86604796-86604799 on Build GRCh38
rs924812236 Chr.2: 86604713-86604713 on Build GRCh38
rs774203865 Chr.2: 86604647-86604647 on Build GRCh38
rs774155968 Chr.2: 86604726-86604726 on Build GRCh38
rs760589907 Chr.2: 86604786-86604786 on Build GRCh38
rs768214868 Chr.2: 86604721-86604721 on Build GRCh38
rs749749696 Chr.2: 86604851-86604851 on Build GRCh38
rs756620778 Chr.2: 86604689-86604689 on Build GRCh38
rs757585213 Chr.2: 86604660-86604660 on Build GRCh38
rs764111646 Chr.2: 86604812-86604812 on Build GRCh38
rs750784881 Chr.2: 86604841-86604841 on Build GRCh38
rs751811518 Chr.2: 86604813-86604813 on Build GRCh38
rs780710494 Chr.2: 86604846-86604846 on Build GRCh38
rs746404741 Chr.2: 86604683-86604683 on Build GRCh38
rs1025555780 Chr.2: 86604799-86604799 on Build GRCh38
rs763038893 Chr.2: 86604800-86604800 on Build GRCh38
rs754553526 Chr.2: 86604853-86604853 on Build GRCh38
rs773026542 Chr.2: 86604779-86604779 on Build GRCh38
rs888098442 Chr.2: 86604662-86604662 on Build GRCh38
rs950323237 Chr.2: 86604678-86604678 on Build GRCh38
rs751723340 Chr.2: 86604654-86604654 on Build GRCh38
rs779767603 Chr.2: 86604708-86604708 on Build GRCh38
rs757472476 Chr.2: 86604818-86604818 on Build GRCh38
rs564118278 Chr.2: 86604731-86604731 on Build GRCh38
rs373564785 Chr.2: 86604681-86604681 on Build GRCh38
rs766338643 Chr.2: 86604791-86604791 on Build GRCh38
rs375660475 Chr.2: 86604755-86604755 on Build GRCh38
rs767823059 Chr.2: 86604830-86604830 on Build GRCh38
rs745639713 Chr.2: 86604720-86604720 on Build GRCh38
rs896869587 Chr.2: 86604772-86604772 on Build GRCh38
rs748802871 Chr.2: 86604718-86604718 on Build GRCh38

View Full Product Details