Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241562209-241562482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOK
Entrez Gene ID
666
Gene Name
BOK, BCL2 family apoptosis regulator
Gene Aliases
BCL2L9, BOKL
UniGene
Hs.293753
Gene Chromosome Location
Chr.2: 241558731-241574138 on Build GRCh38
Transcripts
NM_032515.4 XM_011511696.2 XM_011511697.2 XM_017004775.1 XM_017004776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5862224 c.241C>T p.R81W Chr.2: 241562368-241562368 on Build GRCh38
69839 c.275T>C p.L92P Chr.2: 241562402-241562402 on Build GRCh38
6344136 c.301G>T p.V101L Chr.2: 241562428-241562428 on Build GRCh38
4093107 c.315G>A p.A105A Chr.2: 241562442-241562442 on Build GRCh38
3839104 c.271C>T p.Q91* Chr.2: 241562398-241562398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145752883 Chr.2: 241562389-241562389 on Build GRCh38
rs754152649 Chr.2: 241562345-241562345 on Build GRCh38
rs987721145 Chr.2: 241562372-241562372 on Build GRCh38
rs541674847 Chr.2: 241562335-241562335 on Build GRCh38
rs376254139 Chr.2: 241562234-241562234 on Build GRCh38
rs1028763454 Chr.2: 241562291-241562291 on Build GRCh38
rs1002544684 Chr.2: 241562460-241562460 on Build GRCh38
rs376688053 Chr.2: 241562430-241562430 on Build GRCh38
rs200833733 Chr.2: 241562312-241562312 on Build GRCh38
rs116907701 Chr.2: 241562459-241562459 on Build GRCh38
rs768195034 Chr.2: 241562387-241562387 on Build GRCh38
rs369830668 Chr.2: 241562311-241562311 on Build GRCh38
rs777155155 Chr.2: 241562395-241562395 on Build GRCh38
rs757085403 Chr.2: 241562369-241562369 on Build GRCh38
rs529643299 Chr.2: 241562367-241562367 on Build GRCh38
rs773760888 Chr.2: 241562330-241562330 on Build GRCh38
rs762991733 Chr.2: 241562417-241562417 on Build GRCh38
rs370022649 Chr.2: 241562442-241562442 on Build GRCh38
rs989198073 Chr.2: 241562304-241562304 on Build GRCh38
rs148098112 Chr.2: 241562398-241562398 on Build GRCh38
rs943634937 Chr.2: 241562383-241562383 on Build GRCh38
rs765381402 Chr.2: 241562399-241562399 on Build GRCh38
rs146987702 Chr.2: 241562368-241562368 on Build GRCh38
rs772465016 Chr.2: 241562321-241562321 on Build GRCh38
rs759867602 Chr.2: 241562350-241562350 on Build GRCh38
rs750227285 Chr.2: 241562441-241562441 on Build GRCh38
rs377356735 Chr.2: 241562388-241562388 on Build GRCh38
rs774889642 Chr.2: 241562314-241562314 on Build GRCh38
rs34395510 Chr.2: 241562376-241562377 on Build GRCh38
rs201382143 Chr.2: 241562393-241562393 on Build GRCh38
rs758501848 Chr.2: 241562362-241562362 on Build GRCh38
rs778596750 Chr.2: 241562452-241562452 on Build GRCh38
rs748946523 Chr.2: 241562445-241562445 on Build GRCh38
rs771523960 Chr.2: 241562394-241562394 on Build GRCh38
rs193087751 Chr.2: 241562351-241562351 on Build GRCh38
rs140246126 Chr.2: 241562451-241562451 on Build GRCh38
rs372094266 Chr.2: 241562376-241562376 on Build GRCh38
rs746848417 Chr.2: 241562305-241562305 on Build GRCh38
rs886557259 Chr.2: 241562418-241562418 on Build GRCh38
rs199763136 Chr.2: 241562377-241562377 on Build GRCh38
rs145137590 Chr.2: 241562413-241562413 on Build GRCh38
rs74003741 Chr.2: 241562436-241562436 on Build GRCh38
rs974659210 Chr.2: 241562288-241562288 on Build GRCh38
rs142111641 Chr.2: 241562420-241562420 on Build GRCh38
rs200898291 Chr.2: 241562437-241562437 on Build GRCh38
rs766826365 Chr.2: 241562340-241562340 on Build GRCh38
rs544022176 Chr.2: 241562276-241562276 on Build GRCh38
rs748812814 Chr.2: 241562384-241562384 on Build GRCh38
rs559106129 Chr.2: 241562356-241562356 on Build GRCh38

View Full Product Details