Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41220641-41220894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021040 c.153G>A p.T51T Chr.19: 41220703-41220703 on Build GRCh38
259589 c.128A>C p.N43T Chr.19: 41220678-41220678 on Build GRCh38
7201261 c.166T>G p.C56G Chr.19: 41220716-41220716 on Build GRCh38
3226719 c.148C>T p.L50F Chr.19: 41220698-41220698 on Build GRCh38
4991300 c.303G>C p.Q101H Chr.19: 41220853-41220853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995493128 Chr.19: 41220741-41220741 on Build GRCh38
rs776776986 Chr.19: 41220786-41220786 on Build GRCh38
rs752231721 Chr.19: 41220820-41220820 on Build GRCh38
rs780211796 Chr.19: 41220774-41220774 on Build GRCh38
rs766871347 Chr.19: 41220811-41220811 on Build GRCh38
rs143071942 Chr.19: 41220762-41220762 on Build GRCh38
rs749268016 Chr.19: 41220672-41220672 on Build GRCh38
rs765631388 Chr.19: 41220710-41220710 on Build GRCh38
rs777529927 Chr.19: 41220670-41220670 on Build GRCh38
rs778793836 Chr.19: 41220871-41220871 on Build GRCh38
rs146598679 Chr.19: 41220745-41220745 on Build GRCh38
rs550455192 Chr.19: 41220739-41220739 on Build GRCh38
rs372605787 Chr.19: 41220787-41220787 on Build GRCh38
rs768561674 Chr.19: 41220784-41220784 on Build GRCh38
rs760376383 Chr.19: 41220832-41220832 on Build GRCh38
rs770570685 Chr.19: 41220680-41220680 on Build GRCh38
rs201794937 Chr.19: 41220872-41220872 on Build GRCh38
rs763716654 Chr.19: 41220837-41220837 on Build GRCh38
rs140935892 Chr.19: 41220754-41220754 on Build GRCh38
rs755527465 Chr.19: 41220744-41220744 on Build GRCh38
rs575141970 Chr.19: 41220737-41220737 on Build GRCh38
rs55767963 Chr.19: 41220703-41220703 on Build GRCh38
rs55690306 Chr.19: 41220772-41220772 on Build GRCh38
rs753379207 Chr.19: 41220865-41220865 on Build GRCh38
rs747279308 Chr.19: 41220783-41220783 on Build GRCh38
rs369684238 Chr.19: 41220698-41220698 on Build GRCh38
rs747638130 Chr.19: 41220663-41220663 on Build GRCh38
rs775741490 Chr.19: 41220696-41220696 on Build GRCh38
rs774132867 Chr.19: 41220692-41220692 on Build GRCh38
rs757210880 Chr.19: 41220760-41220760 on Build GRCh38
rs200868176 Chr.19: 41220722-41220722 on Build GRCh38
rs778883675 Chr.19: 41220761-41220761 on Build GRCh38
rs962997099 Chr.19: 41220785-41220785 on Build GRCh38
rs144418422 Chr.19: 41220728-41220728 on Build GRCh38
rs781049505 Chr.19: 41220662-41220662 on Build GRCh38
rs756813396 Chr.19: 41220870-41220870 on Build GRCh38
rs568179015 Chr.19: 41220702-41220702 on Build GRCh38
rs763368586 Chr.19: 41220800-41220800 on Build GRCh38
rs755688015 Chr.19: 41220664-41220664 on Build GRCh38
rs200598880 Chr.19: 41220693-41220693 on Build GRCh38
rs750804190 Chr.19: 41220714-41220714 on Build GRCh38
rs773894785 Chr.19: 41220789-41220789 on Build GRCh38
rs374467939 Chr.19: 41220706-41220706 on Build GRCh38
rs758142178 Chr.19: 41220876-41220876 on Build GRCh38

View Full Product Details