Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1390517-1390762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275927 c.776G>A p.R259H Chr.1: 1390547-1390547 on Build GRCh38
5886026 c.762C>T p.I254I Chr.1: 1390561-1390561 on Build GRCh38
5907845 c.781C>T p.H261Y Chr.1: 1390542-1390542 on Build GRCh38
3801550 c.785G>T p.W262L Chr.1: 1390538-1390538 on Build GRCh38
4393566 c.795G>C p.L265F Chr.1: 1390528-1390528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778813125 Chr.1: 1390594-1390595 on Build GRCh38
rs374357946 Chr.1: 1390567-1390567 on Build GRCh38
rs750848109 Chr.1: 1390602-1390602 on Build GRCh38
rs547776483 Chr.1: 1390653-1390653 on Build GRCh38
rs554464405 Chr.1: 1390630-1390630 on Build GRCh38
rs533536015 Chr.1: 1390582-1390582 on Build GRCh38
rs572597413 Chr.1: 1390701-1390701 on Build GRCh38
rs907127996 Chr.1: 1390707-1390707 on Build GRCh38
rs147017832 Chr.1: 1390610-1390610 on Build GRCh38
rs777311236 Chr.1: 1390600-1390600 on Build GRCh38
rs770217611 Chr.1: 1390718-1390718 on Build GRCh38
rs539890537 Chr.1: 1390708-1390708 on Build GRCh38
rs746533386 Chr.1: 1390728-1390729 on Build GRCh38
rs1028413203 Chr.1: 1390618-1390618 on Build GRCh38
rs758135631 Chr.1: 1390719-1390720 on Build GRCh38
rs754815557 Chr.1: 1390575-1390575 on Build GRCh38
rs370399343 Chr.1: 1390550-1390550 on Build GRCh38
rs778379449 Chr.1: 1390580-1390580 on Build GRCh38
rs747498144 Chr.1: 1390603-1390603 on Build GRCh38
rs768917732 Chr.1: 1390712-1390712 on Build GRCh38
rs766973260 Chr.1: 1390665-1390665 on Build GRCh38
rs987930522 Chr.1: 1390675-1390675 on Build GRCh38
rs756591269 Chr.1: 1390730-1390730 on Build GRCh38
rs779990304 Chr.1: 1390548-1390548 on Build GRCh38
rs910258330 Chr.1: 1390597-1390597 on Build GRCh38
rs1037511991 Chr.1: 1390552-1390552 on Build GRCh38
rs754839610 Chr.1: 1390586-1390587 on Build GRCh38
rs775535842 Chr.1: 1390725-1390725 on Build GRCh38
rs756170860 Chr.1: 1390547-1390547 on Build GRCh38
rs192467977 Chr.1: 1390609-1390609 on Build GRCh38
rs368387971 Chr.1: 1390729-1390729 on Build GRCh38
rs770160058 Chr.1: 1390652-1390652 on Build GRCh38
rs376878356 Chr.1: 1390689-1390689 on Build GRCh38
rs1042956489 Chr.1: 1390624-1390624 on Build GRCh38
rs777277194 Chr.1: 1390720-1390720 on Build GRCh38
rs971279476 Chr.1: 1390558-1390558 on Build GRCh38
rs751753821 Chr.1: 1390735-1390735 on Build GRCh38
rs771535127 Chr.1: 1390595-1390595 on Build GRCh38
rs1012871465 Chr.1: 1390549-1390549 on Build GRCh38
rs984384760 Chr.1: 1390593-1390593 on Build GRCh38
rs750356686 Chr.1: 1390543-1390543 on Build GRCh38
rs531354471 Chr.1: 1390620-1390620 on Build GRCh38
rs747862713 Chr.1: 1390718-1390719 on Build GRCh38
rs965668468 Chr.1: 1390670-1390670 on Build GRCh38
rs937950855 Chr.1: 1390713-1390713 on Build GRCh38
rs903897070 Chr.1: 1390634-1390634 on Build GRCh38
rs967796611 Chr.1: 1390699-1390699 on Build GRCh38
rs771498090 Chr.1: 1390608-1390608 on Build GRCh38
rs1056387112 Chr.1: 1390714-1390714 on Build GRCh38
rs776793893 Chr.1: 1390577-1390577 on Build GRCh38

View Full Product Details