Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26774166-26774423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F6
Entrez Gene ID
54978
Gene Name
solute carrier family 35 member F6
Gene Aliases
ANT2BP, C2orf18, TANGO9
UniGene
Hs.1594
Gene Chromosome Location
Chr.2: 26764274-26781231 on Build GRCh38
Transcripts
NM_017877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3731320 c.105C>T p.G35G Chr.2: 26774278-26774278 on Build GRCh38
6467141 c.125A>G p.H42R Chr.2: 26774298-26774298 on Build GRCh38
4444008 c.82G>A p.A28T Chr.2: 26774255-26774255 on Build GRCh38
3580448 c.139C>T p.P47S Chr.2: 26774312-26774312 on Build GRCh38
7295668 c.118A>G p.K40E Chr.2: 26774291-26774291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754754409 Chr.2: 26774267-26774267 on Build GRCh38
rs779195882 Chr.2: 26774282-26774282 on Build GRCh38
rs775769193 Chr.2: 26774212-26774212 on Build GRCh38
rs769422855 Chr.2: 26774337-26774337 on Build GRCh38
rs540611130 Chr.2: 26774375-26774375 on Build GRCh38
rs752527921 Chr.2: 26774363-26774363 on Build GRCh38
rs1038203675 Chr.2: 26774365-26774365 on Build GRCh38
rs746719535 Chr.2: 26774338-26774339 on Build GRCh38
rs776110791 Chr.2: 26774254-26774254 on Build GRCh38
rs774885920 Chr.2: 26774376-26774376 on Build GRCh38
rs373415480 Chr.2: 26774333-26774333 on Build GRCh38
rs529383256 Chr.2: 26774388-26774388 on Build GRCh38
rs757549752 Chr.2: 26774273-26774273 on Build GRCh38
rs761322027 Chr.2: 26774248-26774248 on Build GRCh38
rs369441743 Chr.2: 26774322-26774322 on Build GRCh38
rs564506232 Chr.2: 26774201-26774201 on Build GRCh38
rs933980339 Chr.2: 26774281-26774281 on Build GRCh38
rs769095314 Chr.2: 26774374-26774374 on Build GRCh38
rs772758191 Chr.2: 26774340-26774340 on Build GRCh38
rs776295893 Chr.2: 26774237-26774237 on Build GRCh38
rs768673307 Chr.2: 26774344-26774345 on Build GRCh38
rs754025096 Chr.2: 26774272-26774272 on Build GRCh38
rs747353384 Chr.2: 26774313-26774313 on Build GRCh38
rs776149072 Chr.2: 26774326-26774326 on Build GRCh38
rs766257768 Chr.2: 26774356-26774356 on Build GRCh38
rs987020297 Chr.2: 26774266-26774266 on Build GRCh38
rs761073156 Chr.2: 26774234-26774234 on Build GRCh38
rs764656606 Chr.2: 26774250-26774250 on Build GRCh38
rs143381489 Chr.2: 26774257-26774257 on Build GRCh38
rs764238638 Chr.2: 26774268-26774268 on Build GRCh38
rs189422020 Chr.2: 26774236-26774236 on Build GRCh38
rs762417936 Chr.2: 26774344-26774344 on Build GRCh38
rs375203801 Chr.2: 26774294-26774294 on Build GRCh38
rs369467381 Chr.2: 26774204-26774204 on Build GRCh38
rs544931894 Chr.2: 26774193-26774193 on Build GRCh38
rs746115578 Chr.2: 26774361-26774361 on Build GRCh38
rs937943732 Chr.2: 26774401-26774401 on Build GRCh38
rs772703241 Chr.2: 26774251-26774251 on Build GRCh38
rs751349829 Chr.2: 26774262-26774262 on Build GRCh38
rs144526895 Chr.2: 26774302-26774302 on Build GRCh38
rs979658118 Chr.2: 26774383-26774383 on Build GRCh38
rs375390451 Chr.2: 26774252-26774252 on Build GRCh38
rs370078739 Chr.2: 26774384-26774384 on Build GRCh38
rs373059645 Chr.2: 26774359-26774359 on Build GRCh38
rs762934984 Chr.2: 26774256-26774256 on Build GRCh38
rs763605579 Chr.2: 26774358-26774358 on Build GRCh38
rs745924341 Chr.2: 26774289-26774289 on Build GRCh38

View Full Product Details