Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52403736-52403963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580073 c.1357A>G p.K453E Chr.3: 52403788-52403788 on Build GRCh38
1424460 c.1338C>T p.N446N Chr.3: 52403807-52403807 on Build GRCh38
6925080 c.1251-2A>T p.? Chr.3: 52403896-52403896 on Build GRCh38
5708825 c.1251-1G>A p.? Chr.3: 52403895-52403895 on Build GRCh38
1732101 c.1306C>T p.Q436* Chr.3: 52403839-52403839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915913191 Chr.3: 52403854-52403854 on Build GRCh38
rs757188301 Chr.3: 52403821-52403823 on Build GRCh38
rs772437485 Chr.3: 52403844-52403844 on Build GRCh38
rs145361939 Chr.3: 52403758-52403758 on Build GRCh38
rs184055262 Chr.3: 52403928-52403928 on Build GRCh38
rs761544042 Chr.3: 52403894-52403894 on Build GRCh38
rs202038162 Chr.3: 52403837-52403837 on Build GRCh38
rs750343039 Chr.3: 52403898-52403898 on Build GRCh38
rs77722216 Chr.3: 52403825-52403825 on Build GRCh38
rs772505081 Chr.3: 52403760-52403760 on Build GRCh38
rs756784645 Chr.3: 52403840-52403840 on Build GRCh38
rs779304334 Chr.3: 52403925-52403925 on Build GRCh38
rs762422208 Chr.3: 52403881-52403881 on Build GRCh38
rs371601284 Chr.3: 52403863-52403863 on Build GRCh38
rs756136482 Chr.3: 52403901-52403901 on Build GRCh38
rs866182083 Chr.3: 52403850-52403850 on Build GRCh38
rs781013694 Chr.3: 52403811-52403811 on Build GRCh38
rs758996824 Chr.3: 52403787-52403787 on Build GRCh38
rs754019811 Chr.3: 52403913-52403913 on Build GRCh38
rs763735807 Chr.3: 52403807-52403807 on Build GRCh38
rs764734266 Chr.3: 52403800-52403800 on Build GRCh38
rs755059361 Chr.3: 52403914-52403914 on Build GRCh38
rs767276752 Chr.3: 52403897-52403897 on Build GRCh38
rs115109161 Chr.3: 52403877-52403877 on Build GRCh38
rs374746213 Chr.3: 52403815-52403815 on Build GRCh38
rs763789102 Chr.3: 52403886-52403886 on Build GRCh38
rs771326820 Chr.3: 52403869-52403869 on Build GRCh38
rs766385310 Chr.3: 52403902-52403902 on Build GRCh38
rs587776877 Chr.3: 52403840-52403840 on Build GRCh38
rs758700978 Chr.3: 52403935-52403935 on Build GRCh38
rs375691067 Chr.3: 52403803-52403803 on Build GRCh38
rs762654322 Chr.3: 52403806-52403806 on Build GRCh38
rs587778089 Chr.3: 52403820-52403820 on Build GRCh38
rs368684241 Chr.3: 52403801-52403801 on Build GRCh38
rs749372454 Chr.3: 52403822-52403822 on Build GRCh38
rs751399960 Chr.3: 52403808-52403808 on Build GRCh38
rs780414230 Chr.3: 52403867-52403867 on Build GRCh38
rs769038290 Chr.3: 52403823-52403823 on Build GRCh38
rs773715938 Chr.3: 52403783-52403783 on Build GRCh38
rs924673060 Chr.3: 52403906-52403906 on Build GRCh38
rs773947541 Chr.3: 52403892-52403892 on Build GRCh38
rs756153541 Chr.3: 52403817-52403817 on Build GRCh38
rs981419588 Chr.3: 52403905-52403905 on Build GRCh38
rs761183483 Chr.3: 52403865-52403865 on Build GRCh38
rs773592865 Chr.3: 52403855-52403855 on Build GRCh38

View Full Product Details