Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28737452-28737725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3382207 c.2400A>C p.T800T Chr.8: 28737642-28737642 on Build GRCh38
1623901 c.2334G>T p.W778C Chr.8: 28737576-28737576 on Build GRCh38
310970 c.2417A>G p.H806R Chr.8: 28737659-28737659 on Build GRCh38
6657430 c.2420A>G p.K807R Chr.8: 28737662-28737662 on Build GRCh38
1489219 c.2361C>G p.Y787* Chr.8: 28737603-28737603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35067545 Chr.8: 28737599-28737600 on Build GRCh38
rs540742947 Chr.8: 28737570-28737570 on Build GRCh38
rs754831873 Chr.8: 28737497-28737497 on Build GRCh38
rs751188982 Chr.8: 28737478-28737478 on Build GRCh38
rs369118658 Chr.8: 28737502-28737502 on Build GRCh38
rs774735721 Chr.8: 28737563-28737563 on Build GRCh38
rs779843733 Chr.8: 28737507-28737507 on Build GRCh38
rs148404093 Chr.8: 28737582-28737582 on Build GRCh38
rs375440390 Chr.8: 28737573-28737573 on Build GRCh38
rs973242051 Chr.8: 28737559-28737559 on Build GRCh38
rs371812947 Chr.8: 28737479-28737479 on Build GRCh38
rs758450574 Chr.8: 28737520-28737520 on Build GRCh38
rs144474985 Chr.8: 28737519-28737519 on Build GRCh38
rs769361447 Chr.8: 28737547-28737547 on Build GRCh38
rs781131046 Chr.8: 28737542-28737542 on Build GRCh38
rs369382883 Chr.8: 28737571-28737571 on Build GRCh38
rs983734401 Chr.8: 28737539-28737539 on Build GRCh38
rs375444536 Chr.8: 28737555-28737555 on Build GRCh38
rs777196726 Chr.8: 28737680-28737680 on Build GRCh38
rs530700503 Chr.8: 28737692-28737692 on Build GRCh38
rs748141724 Chr.8: 28737669-28737669 on Build GRCh38
rs751320973 Chr.8: 28737643-28737643 on Build GRCh38
rs992870080 Chr.8: 28737496-28737496 on Build GRCh38
rs190213591 Chr.8: 28737546-28737546 on Build GRCh38
rs376747953 Chr.8: 28737495-28737495 on Build GRCh38
rs756857596 Chr.8: 28737501-28737502 on Build GRCh38
rs181847256 Chr.8: 28737691-28737691 on Build GRCh38
rs7819432 Chr.8: 28737618-28737618 on Build GRCh38
rs756315681 Chr.8: 28737499-28737499 on Build GRCh38
rs373190688 Chr.8: 28737600-28737600 on Build GRCh38
rs768599793 Chr.8: 28737541-28737541 on Build GRCh38
rs532555915 Chr.8: 28737651-28737651 on Build GRCh38
rs148896601 Chr.8: 28737688-28737688 on Build GRCh38
rs759837681 Chr.8: 28737565-28737565 on Build GRCh38
rs762607710 Chr.8: 28737549-28737549 on Build GRCh38
rs779037344 Chr.8: 28737625-28737625 on Build GRCh38
rs756057803 Chr.8: 28737670-28737670 on Build GRCh38
rs780703404 Chr.8: 28737563-28737563 on Build GRCh38
rs544827563 Chr.8: 28737504-28737504 on Build GRCh38

View Full Product Details