Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138198210-138198441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC23
Entrez Gene ID
8697
Gene Name
cell division cycle 23
Gene Aliases
ANAPC8, APC8, CUT23
UniGene
Hs.73625
Gene Chromosome Location
Chr.5: 138187648-138213343 on Build GRCh38
Transcripts
NM_004661.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267826 c.931-3C>A p.? Chr.5: 138198283-138198283 on Build GRCh38
3608905 c.1000T>G p.C334G Chr.5: 138198211-138198211 on Build GRCh38
1433345 c.938A>C p.K313T Chr.5: 138198273-138198273 on Build GRCh38
6293178 c.936G>A p.M312I Chr.5: 138198275-138198275 on Build GRCh38
237166 c.990T>C p.R330R Chr.5: 138198221-138198221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759878459 Chr.5: 138198324-138198324 on Build GRCh38
rs765620800 Chr.5: 138198381-138198381 on Build GRCh38
rs546810317 Chr.5: 138198373-138198374 on Build GRCh38
rs768132194 Chr.5: 138198294-138198294 on Build GRCh38
rs759932217 Chr.5: 138198375-138198375 on Build GRCh38
rs753009798 Chr.5: 138198386-138198386 on Build GRCh38
rs762693491 Chr.5: 138198285-138198285 on Build GRCh38
rs374243034 Chr.5: 138198316-138198316 on Build GRCh38
rs2231476 Chr.5: 138198341-138198341 on Build GRCh38
rs769684363 Chr.5: 138198272-138198272 on Build GRCh38
rs939241978 Chr.5: 138198300-138198300 on Build GRCh38
rs761222789 Chr.5: 138198311-138198311 on Build GRCh38
rs758651498 Chr.5: 138198399-138198399 on Build GRCh38
rs928658107 Chr.5: 138198297-138198297 on Build GRCh38
rs899542253 Chr.5: 138198345-138198345 on Build GRCh38
rs1050723718 Chr.5: 138198365-138198365 on Build GRCh38
rs775389485 Chr.5: 138198277-138198277 on Build GRCh38
rs370310516 Chr.5: 138198312-138198312 on Build GRCh38
rs186304061 Chr.5: 138198319-138198319 on Build GRCh38
rs781263048 Chr.5: 138198245-138198245 on Build GRCh38
rs997020695 Chr.5: 138198352-138198352 on Build GRCh38
rs745709624 Chr.5: 138198270-138198270 on Build GRCh38
rs377161370 Chr.5: 138198303-138198303 on Build GRCh38
rs1051041742 Chr.5: 138198404-138198404 on Build GRCh38

View Full Product Details