Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30716447-30716718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019864.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199549 c.2296-1G>T p.? Chr.12: 30716677-30716677 on Build GRCh38
7260473 c.2411A>G p.Q804R Chr.12: 30716561-30716561 on Build GRCh38
5739602 c.2461A>T p.T821S Chr.12: 30716511-30716511 on Build GRCh38
1188462 c.2378G>T p.S793I Chr.12: 30716594-30716594 on Build GRCh38
5699067 c.2424A>T p.P808P Chr.12: 30716548-30716548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756534335 Chr.12: 30716645-30716645 on Build GRCh38
rs907865181 Chr.12: 30716542-30716542 on Build GRCh38
rs754335721 Chr.12: 30716656-30716656 on Build GRCh38
rs779041426 Chr.12: 30716576-30716576 on Build GRCh38
rs748327785 Chr.12: 30716579-30716579 on Build GRCh38
rs762410762 Chr.12: 30716614-30716614 on Build GRCh38
rs755040329 Chr.12: 30716659-30716659 on Build GRCh38
rs368121707 Chr.12: 30716594-30716594 on Build GRCh38
rs769603497 Chr.12: 30716681-30716681 on Build GRCh38
rs777961163 Chr.12: 30716674-30716674 on Build GRCh38
rs766843873 Chr.12: 30716655-30716655 on Build GRCh38
rs750981136 Chr.12: 30716544-30716544 on Build GRCh38
rs988475691 Chr.12: 30716471-30716471 on Build GRCh38
rs141863140 Chr.12: 30716662-30716662 on Build GRCh38
rs781648821 Chr.12: 30716541-30716541 on Build GRCh38
rs780163021 Chr.12: 30716688-30716688 on Build GRCh38
rs759654328 Chr.12: 30716615-30716615 on Build GRCh38
rs751194223 Chr.12: 30716536-30716536 on Build GRCh38
rs1031788472 Chr.12: 30716667-30716667 on Build GRCh38
rs780607029 Chr.12: 30716558-30716558 on Build GRCh38
rs768757301 Chr.12: 30716562-30716562 on Build GRCh38
rs1046652876 Chr.12: 30716565-30716565 on Build GRCh38
rs371982877 Chr.12: 30716660-30716660 on Build GRCh38
rs148110140 Chr.12: 30716637-30716637 on Build GRCh38
rs944818832 Chr.12: 30716568-30716568 on Build GRCh38
rs745716115 Chr.12: 30716679-30716679 on Build GRCh38
rs983860024 Chr.12: 30716528-30716528 on Build GRCh38
rs774172019 Chr.12: 30716621-30716621 on Build GRCh38
rs763706404 Chr.12: 30716522-30716522 on Build GRCh38
rs764811341 Chr.12: 30716492-30716492 on Build GRCh38
rs749384115 Chr.12: 30716560-30716560 on Build GRCh38
rs375341933 Chr.12: 30716554-30716554 on Build GRCh38
rs761501370 Chr.12: 30716636-30716636 on Build GRCh38
rs912863532 Chr.12: 30716495-30716495 on Build GRCh38
rs534456727 Chr.12: 30716668-30716668 on Build GRCh38
rs769682193 Chr.12: 30716600-30716600 on Build GRCh38
rs751618967 Chr.12: 30716610-30716610 on Build GRCh38
rs752219767 Chr.12: 30716502-30716502 on Build GRCh38
rs770731505 Chr.12: 30716580-30716580 on Build GRCh38
rs999781647 Chr.12: 30716663-30716663 on Build GRCh38
rs757729711 Chr.12: 30716538-30716538 on Build GRCh38
rs763620955 Chr.12: 30716620-30716620 on Build GRCh38

View Full Product Details