Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77605769-77606022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL13
Entrez Gene ID
10563
Gene Name
C-X-C motif chemokine ligand 13
Gene Aliases
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13
UniGene
Hs.100431
Gene Chromosome Location
Chr.4: 77511753-77611834 on Build GRCh38
Transcripts
NM_006419.2 XM_006714063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4529299 c.15G>A p.S5S Chr.4: 77605880-77605880 on Build GRCh38
420515 c.30C>T p.L10L Chr.4: 77605895-77605895 on Build GRCh38
1057756 c.14C>T p.S5L Chr.4: 77605879-77605879 on Build GRCh38
5750164 c.8T>A p.F3Y Chr.4: 77605873-77605873 on Build GRCh38
222922 c.20C>T p.S7F Chr.4: 77605885-77605885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767463762 Chr.4: 77605920-77605920 on Build GRCh38
rs2270858 Chr.4: 77605989-77605989 on Build GRCh38
rs573522595 Chr.4: 77605797-77605797 on Build GRCh38
rs747589194 Chr.4: 77605857-77605857 on Build GRCh38
rs766030835 Chr.4: 77605925-77605925 on Build GRCh38
rs201522414 Chr.4: 77605960-77605960 on Build GRCh38
rs528808494 Chr.4: 77605981-77605981 on Build GRCh38
rs760469717 Chr.4: 77605924-77605924 on Build GRCh38
rs781034959 Chr.4: 77605955-77605958 on Build GRCh38
rs972581246 Chr.4: 77605972-77605972 on Build GRCh38
rs746262587 Chr.4: 77605879-77605879 on Build GRCh38
rs200616061 Chr.4: 77605941-77605941 on Build GRCh38
rs201339877 Chr.4: 77605888-77605888 on Build GRCh38
rs1053385407 Chr.4: 77605805-77605805 on Build GRCh38
rs758548525 Chr.4: 77605940-77605940 on Build GRCh38
rs777187615 Chr.4: 77605858-77605858 on Build GRCh38
rs893520712 Chr.4: 77605813-77605813 on Build GRCh38
rs777705572 Chr.4: 77605850-77605850 on Build GRCh38
rs906390267 Chr.4: 77605913-77605913 on Build GRCh38
rs919419727 Chr.4: 77605979-77605979 on Build GRCh38
rs762496301 Chr.4: 77605890-77605890 on Build GRCh38
rs761786107 Chr.4: 77605914-77605914 on Build GRCh38
rs763725491 Chr.4: 77605893-77605893 on Build GRCh38
rs755855539 Chr.4: 77605815-77605815 on Build GRCh38
rs757369386 Chr.4: 77605948-77605948 on Build GRCh38
rs779229051 Chr.4: 77605844-77605844 on Build GRCh38
rs769983573 Chr.4: 77605880-77605880 on Build GRCh38
rs993158214 Chr.4: 77605985-77605985 on Build GRCh38
rs201605324 Chr.4: 77605943-77605943 on Build GRCh38
rs748252694 Chr.4: 77605849-77605849 on Build GRCh38
rs773818843 Chr.4: 77605912-77605912 on Build GRCh38
rs1019839507 Chr.4: 77605836-77605836 on Build GRCh38
rs994312843 Chr.4: 77605877-77605877 on Build GRCh38
rs549377637 Chr.4: 77605942-77605942 on Build GRCh38
rs202112173 Chr.4: 77605923-77605923 on Build GRCh38

View Full Product Details