Gene Transcripts
- Gene Symbol
- Acacb
- Entrez Gene ID
- 100705
- Gene Name
- acetyl-Coenzyme A carboxylase beta
- Gene Aliases
- AI597064, AW743042, Acc2, Accb
- Chromosome Location
- Chr. 5: 114146318 - 114250761 on Build GRCm38
- UniGene ID
- Mm.81793
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133904.2 |
NP_598665.2
|
11 |
1824 |
|
GenBank mRNA |
AY451394.1 |
- |
11 |
1824 |
|
View Full Product Details