Gene Transcripts
- Gene Symbol
- Srr
- Entrez Gene ID
- 27364
- Gene Name
- serine racemase
- Gene Aliases
- Srs
- Chromosome Location
- Chr. 11: 74905047 - 74925798 on Build GRCm38
- UniGene ID
- Mm.131443
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK002636.1 |
- |
|
1243 |
|
- Gene Symbol
- Tsr1
- Entrez Gene ID
- 104662
- Gene Name
- TSR1 20S rRNA accumulation
- Gene Aliases
- AU040765, AW550801, mKIAA1401
- Chromosome Location
- Chr. 11: 74898080 - 74909340 on Build GRCm38
- UniGene ID
- Mm.220843
- Species
- Mus musculus
View Full Product Details