Gene Transcripts

Gene Symbol
Pelo
Entrez Gene ID
105083
Gene Name
pelota homolog (Drosophila)
Gene Aliases
AA409897, AI852218
Chromosome Location
Chr. 13: 115088355 - 115090158 on Build GRCm38
UniGene ID
Mm.486295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134058.3 NP_598819.2 2 1267
GenBank mRNA AF148638.1 - 1328
AI854576.1 - 1 318
AK077286.1 - 754
AK146382.1 - 2 1297
AK168228.1 - 2 1297
AK170635.1 - 2 1267
BB767039.1 - 1 242
BC050209.1 - 2 1180 6431710
BC057160.1 - 2 1068 5349483

View Full Product Details