Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
110172
Gene Name
solute carrier family 35, member B1
Gene Aliases
Ugalt2
Chromosome Location
Chr. 11: 95384891 - 95391776 on Build GRCm38
UniGene ID
Mm.4593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016752.1 NP_058032.1 7 811
GenBank mRNA AK002961.1 - 7 805
AK007711.1 - 7 817
AK160124.1 - 7 878
BC002029.1 - 7 792 3592141
D87990.1 - 7 811

View Full Product Details