Gene Transcripts

Gene Symbol
Fcer2a
Entrez Gene ID
14128
Gene Name
Fc receptor, IgE, low affinity II, alpha polypeptide
Gene Aliases
CD23, Fce2, Fcer2, Ly-42
Chromosome Location
Chr. 8: 3680726 - 3694174 on Build GRCm38
UniGene ID
Mm.1233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253737.1 NP_001240666.1 11 957
NM_001253739.1 NP_001240668.1 11 968
NM_001253743.1 NP_001240672.1 12 1017
NM_001253745.1 NP_001240674.1 10 894
NM_001253746.1 NP_001240675.1 10 894
NM_001253747.1 NP_001240676.1 11 978
NM_013517.4 NP_038545.1 12 1098
GenBank mRNA AK156654.1 - 12 1101
AK172591.1 - 12 1101
AY069980.1 - 10 860
AY069981.1 - 10 860
BC119164.1 - 11 965 8733913
BC119190.1 - 11 965 8733939
M34163.1 - 12 1100
M99371.1 - 12 1090
S67072.1 - 11 949
S67073.1 - 10 829
S67074.1 - 11 868
X64223.1 - 11 957
X64224.1 - 11 968

View Full Product Details