Gene Transcripts

Gene Symbol
Mcoln3
Entrez Gene ID
171166
Gene Name
mucolipin 3
Gene Aliases
6720490O21Rik, TRPML3, Va
Chromosome Location
Chr. 3: 146093277 - 146141808 on Build GRCm38
UniGene ID
Mm.114683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134160.1 NP_598921.1 12 1560
GenBank mRNA AF475086.1 - 12 1560
AK030819.1 - 12 1680
AK033008.1 - 12 1674
AK035029.1 - 12 1681
AY083531.1 - 12 1558
BC106856.2 - 12 1560 40047039
BC106857.2 - 12 1560 40047042

View Full Product Details